Skip to Content
Merck
All Photos(2)

Key Documents

1.00930

Supelco

Diethyl ether

≥98% (GC), contains 2% ethanol as stabilizer, suitable for UV/Vis spectroscopy, Uvasol®

Synonym(s):

Ethyl ether, Ether, Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-467-2
NACRES:
NA.03
Assay:
≥98% (GC)
technique(s):
UV/Vis spectroscopy: suitable
bp:
34.6 °C (lit.)
vapor pressure:
587 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Diethyl ether, for spectroscopy Uvasol®

vapor density

2.6 (vs air)

Quality Level

vapor pressure

587 hPa ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

320 °F

potency

1215 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

contains

2% ethanol as stabilizer

expl. lim.

36.5 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.0005% Peroxide
≤0.03% Water

evapn. residue

≤0.0003%

color

APHA: ≤10

transmittance

220 nm, ≥30%
235 nm, ≥55%
250 nm, ≥80%
270 nm, ≥90%
300 nm, ≥98%

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

transition temp

flash point -40 °C

density

0.706 g/mL at 25 °C (lit.)

UV absorption

λ: 220 nm Amax: ≤0.52
λ: 235 nm Amax: ≤0.26
λ: 250 nm Amax: ≤0.10
λ: 270 nm Amax: ≤0.05
λ: 300 nm Amax: ≤0.01

storage temp.

15-25°C

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service