Skip to Content
Merck
All Photos(1)

Key Documents

PX0425

Supelco

Petroleum Ether, BP

Meets ACS Specifications GR ACS

Synonym(s):

Petroleum ether, Ligroine,Petroleum Spirit, Benzine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352401
grade:
ACS reagent
bp:
30-50 °C/1013 hPa

grade

ACS reagent

Quality Level

form

liquid

impurities

≤0.01% Water (H2O)

evapn. residue

≤0.001%

color

APHA: ≤10

bp

30-50 °C/1013 hPa

transition temp

flash point <-30 °C

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications GR ACS
PX0425P-4 is packaged in poly coated bottles

Analysis Note

AcidityTo pass testBoiling range35-60°CColor (APHA)10 maxEvaporation residue0.001% maxFormClear liquidWater (H2O)0.01% max

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 1 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-22.0 °F

Flash Point(C)

< -30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service