Skip to Content
Merck
All Photos(1)

Key Documents

1.01707

Sigma-Aldrich

Calcium carbonate

6700002749, Technipur®

Synonym(s):

Calcite, Limestone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

pH

9.5-10.5 (20 °C, 100 g/L in H2O)

mp

825 °C

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium carbonate (also called whiting) is the most widely used additive in plastic and elastomeric applications due to its wide availability, easy processing to specific particle sizes and its compatibility with a wide range of polymer types.[1][2][3]

Application

Calcium carbonate can be used as an inorganic filler in printed electronic applications and the plastics industry to improve the mechanical stability, flexibility, rheological and optical properties of the synthesized composite materials. In polymer formulations, CaCO3 may be used to formulate several thermoplastics (PVC pipes and floor tiles) and thermosets such as polyesters.[1][4][3]

It can also be used as a versatile additive in a wide range of applications such as adhesives, sealants, decorating filler, ceramic glaze, plastics, rubber, coatings, inks, etc.[5]

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The application of basalt in the manufacturing of ceramic glazes
Prstic, A, et al.
Journal of Mining and Metallurgy A: Mining, 43(1), 53-60 (2007)
Filler-nanocellulose substrate for printed electronics: experiments and model approach to structure and conductivity
Penttila, A, et al.
Cellulose, 20(3), 1413-1424 (2013)
Effect of calcium carbonate particle size and content on polyamide 6 processing and properties
Bonse BC and Molina LM
AIP Conference Proceedings, 1779(1), 030019-030019 (2016)
Rheological and optical properties of polypropylene filled with synthetic silicates and calcium carbonate
Upadhyay RD and Kale DD
Journal of Polymer Research, 8(3), 175-181 (2001)
Polymer Matrix Composites: Fundamentals
Beaumont PWR, et al.
Comprehensive Composite Materials II (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service