![Calcium Carbonate NIST® SRM® 915c](/deepweb/assets/sigmaaldrich/product/structures/142/105/167d796b-1988-4b48-a07d-9cbe9fa09fba/640/167d796b-1988-4b48-a07d-9cbe9fa09fba.png)
Calcium carbonate
![Calcium Carbonate NIST® SRM® 915c](/deepweb/assets/sigmaaldrich/product/structures/142/105/167d796b-1988-4b48-a07d-9cbe9fa09fba/640/167d796b-1988-4b48-a07d-9cbe9fa09fba.png)
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 25
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2