Skip to Content
Merck
All Photos(3)

Key Documents

1.00985

Supelco

Isobutanol

(isobutyl alcohol) EMPLURA®

Synonym(s):

Isobutanol, Isobutylalcohol, 2-Methyl-1-propanol, Iso-Butanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
MDL number:
UNSPSC Code:
12191502
EC Index Number:
201-148-0
NACRES:
NA.21
bp:
106-108 °C/1013 hPa
vapor pressure:
12 hPa ( 20 °C)

vapor pressure

12 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥98.5% (GC)

form

liquid

autoignition temp.

430 °C

potency

>2830 mg/kg LD50, oral (Rat)
2460 mg/kg LD50, skin (Rat)

expl. lim.

1.5-12 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.005% Free acid (as C3H7COOH)
≤0.02% Acetone (GC)
≤0.03% Isobutyraldehyde (GC)
≤0.05% Water

pH

7 (20 °C, 80 g/L in H2O)

bp

106-108 °C/1013 hPa

mp

-108 °C

transition temp

flash point 28 °C

solubility

85 g/L

density

0.802 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Application of Fourier-Transform Infrared Spectroscopy for the Assessment of Wine Spoilage Indicators: This study showcases the use of isobutanol as a standard in assessing wine quality through spectroscopic methods, highlighting its role in ensuring the quality and safety of food and beverages (Teixeira Dos Santos et al., 2024).

  • Energy-Saving Electrochemical Hydrogen Production Coupled with Biomass-Derived Isobutanol Upgrading: Focuses on integrating isobutanol production from biomass with hydrogen production processes, illustrating its application in enhancing energy efficiency and renewable energy technologies (Du et al., 2024).

  • Effects of Core Size and Surfactant Choice on Fluid Saturation Development in Surfactant/Polymer Corefloods: Although indirectly related, this research can incorporate isobutanol as a component in formulations to optimize the extraction processes in the oil and gas industry, showcasing its role in industrial applications (Rovelli et al., 2024).

Analysis Note

Purity (GC): ≥ 98.5 %
Identity (IR): conforms
Free acid (as C₃H₇COOH): ≤ 0.005 %
Density (d 20 °C/ 4 °C): 0.801 - 0.802
Acetone (GC): ≤ 0.02 %
Isobutyraldehyde (GC): ≤ 0.03 %
Water: ≤ 0.05 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service