Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diethyl ether
Synonym(s):
Ether, Ethyl ether, Diethyl ether
Linear Formula:
(CH
3
CH
2
)
2
O
CAS Number:
60-29-7
Molecular Weight:
74.12
EC Number:
200-467-2
Products
Product Category
solvents (21)
Brand
SAFC (1)
Sigma-Aldrich (11)
Supelco (10)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (3)
Anhydrous (3)
Puriss (2)
Puriss P.A. (2)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
673811
Diethyl ether,
≥99.0%, anhydrous, ACS reagent, contains BHT as inhibitor
Expand
View Pricing
309966
Diethyl ether,
≥99.9%, suitable for HPLC, inhibitor-free
Expand
View Pricing
346136
Diethyl ether,
≥99.0%, anhydrous, ACS reagent, contains BHT as inhibitor
Expand
View Pricing
1.00921
Diethyl ether,
≥99.7% (GC), ACS reagent, contains 5-10 ppm 2, 6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer, reag. Ph. Eur., reag. ISO, EMSURE
®
Expand
View Pricing
EX0190
Diethyl ether,
≥99.0% (GC), anhydrous, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Expand
View Pricing
32203
Diethyl ether,
≥99.8% (GC), ACS reagent, puriss. p.a., contains BHT as inhibitor, reag. Ph. Eur., reag. ISO, USP/NF
Expand
View Pricing
179272
Diethyl ether,
≥97.5%, contains BHT as stabilizer
Expand
View Pricing
676845
Diethyl ether,
≥98.0%, ACS reagent, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Expand
View Pricing
346144
Diethyl ether,
≥98%, reagent grade, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Expand
View Pricing
24004
Diethyl ether,
≥99.5% (GC), puriss, contains ~5 mg/L 2, 6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP
Expand
View Pricing
5.89589
Diethyl ether,
≥99.7%, anhydrous, contains 1 ppm BHT as inhibitor
Expand
View Pricing
1.00923
Diethyl ether,
≥99% (GC), EMPLURA
®
Expand
View Pricing
EX0185
Diethyl ether,
≥98.0% ethanol basis (diethyl ether), Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Expand
View Pricing
1.00926
Diethyl ether,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP
Expand
View Pricing
296082
Diethyl ether,
≥99.7%, anhydrous, contains 1 ppm BHT as inhibitor
Expand
View Pricing
1.00931
Diethyl ether,
≥98% (GC), suitable for gas chromatography ECD and FID, SupraSolv
®
Expand
View Pricing
1.07026
Diethyl ether,
≥99.5% (GC), EMPARTA
®
ACS reagent, contains 5-10 ppm 2, 6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer
Expand
View Pricing
PHR1666
Diethyl ether,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
91238
Diethyl ether,
analytical standard
Expand
View Pricing
1.00930
Diethyl ether,
≥98% (GC), contains 2% ethanol as stabilizer, suitable for UV/Vis spectroscopy, Uvasol
®
Expand
View Pricing
Page 1 of 2
Page 1 of 2