Skip to Content
Merck
All Photos(1)

Key Documents

1.00842

Millipore

Esculin

suitable for microbiology

Synonym(s):

Esculin, 6,7-Dihydroxycumarin-6ß-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O9 · 1.5H2O
CAS Number:
Molecular Weight:
367.31
MDL number:
UNSPSC Code:
41106212
EC Index Number:
208-517-5
NACRES:
NA.74

Pricing and availability is not currently available.

Assay

≥98.0% (HPLC)

Quality Level

form

solid

mp

212 °C (decomposition)

bulk density

200 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2cc3c([o][c](cc3)=O)cc2O.O

InChI

1S/C15H16O9.H2O/c16-5-10-12(19)13(20)14(21)15(24-10)23-9-3-6-1-2-11(18)22-8(6)4-7(9)17;/h1-4,10,12-17,19-21H,5H2;1H2/t10-,12-,13+,14-,15-;/m1./s1

InChI key

CQYPGSKIFJFVDQ-QWFKVUSTSA-N

Analysis Note

Purity (HPLC): ≥ 98.0 %
Identity (IR-spectrum): conforms
Spec. rotation (α 20/D; 2 %; dioxan/water (1:1); calc. on anhydrous substance): -87 - -84 °
Heavy metals (as Pb): ≤ 0.004 %
Fraxin (HPLC): ≤ 0.5 %
Sulfated ash: ≤ 0.1 %
Water: 6.5 - 8.0 %
Suitability for microbiology: conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service