Skip to Content
Merck
All Photos(1)

Key Documents

07-823

Sigma-Aldrich

Anti-phospho eIF4E (Ser209) Antibody

Upstate®, from rabbit

Synonym(s):

eukaryotic translation initiation factor 4E, CBP, EIF4EL1, EIF4F

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
11

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (100% homologous), mouse (100% homologous), rabbit (100% homologous)

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer209)

Gene Information

human ... EIF4E(1977)

Specificity

Recognizes eIF4E phosphorylated on Ser209.

Immunogen

KLH conjugated synthetic peptide corresponding to amino acids surrounding Ser209 of human eIF4E. Sequence conserved in mouse and rat.

Application

Anti-phospho eIF4E (Ser209) Antibody is an antibody against phospho eIF4E (Ser209) for use in WB.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

RNA Binding Protein (RBP)

Quality

Routinely evaluated by immunoblot.

Target description

25 kDa

Physical form

ImmunoAffinity Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylation of eukaryotic initiation factor 4E (eIF4E) at Ser209 is not required for protein synthesis in vitro and in vivo
McKendrick, L, et al
European Journal of Biochemistry, 268, 5375-5385 (2001)
B Joshi et al.
The Journal of biological chemistry, 270(24), 14597-14603 (1995-06-16)
Initiation factor 4E (eIF-4E) binds to the m7GTP-containing cap of eukaryotic mRNA and facilitates the entry of mRNA into the initiation cycle of protein synthesis. eIF-4E is a phosphoprotein, and the phosphorylated form binds to mRNA caps 3-4-fold more tightly
A J Waskiewicz et al.
Molecular and cellular biology, 19(3), 1871-1880 (1999-02-18)
Eukaryotic translation initiation factor 4E (eIF4E) binds to the mRNA 5' cap and brings the mRNA into a complex with other protein synthesis initiation factors and ribosomes. The activity of mammalian eIF4E is important for the translation of capped mRNAs
Phosphorylation of eukaryotic initiation factor 4E markedly reduces its affinity for capped mRNA
Scheper, Gert C, et al
The Journal of Biological Chemistry, 277, 3303-3309 (2002)
Joanna Zuberek et al.
Biochemistry, 43(18), 5370-5379 (2004-05-05)
Eukaryotic translation initiation factor 4E (eIF4E) is essential for efficient protein synthesis in cap-dependent translation. The protein specifically binds the cap structure at the mRNA 5' terminus and facilitates the assembly of the mRNA with other initiation factors and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service