Skip to Content
Merck
All Photos(1)

Key Documents

AB1557P

Sigma-Aldrich

Anti-NMDAR2B Antibody

Chemicon®, from rabbit

Synonym(s):

N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 2B, N-methyl-D-aspartate receptor subunit 3, glutamate receptor subunit epsilon-2, glutamate receptor. ionotropic, N-methyl D-aspartate 2B, Glutamate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

General description

NMDA receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long term potentiation, an activity dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D). GRIN2B may be a candidate gene for the neurodegenerative disorder dentato-rubro-pallidoluysian atrophy (DRPLA).
Properties of NMDAR include modulation by glycine, inhibition by Zn2+, voltage dependent Mg2+ blockade and high Ca2+ permeability. The involvement of NMDAR in the CNS has become a focus area for neurodegenerative diseases such as Alzheimer′s disease and also epilepsy and ischemic neuronal cell death.

Specificity

NMDAR2B. Strong and specific immunolabeling of the 180 kDa NMDAR2B subunit of the NMDA receptor. No reactivity to NMDAR2A or NMDAR2C by Western blot. Immunolabeling has been blocked by preadsorption of the antibody with the C-terminal fusion protein that was used to generate the antibody.

Immunogen

C-Terminal Fusion Protein of the NMDAR2B (30 kDa).
Epitope: C-terminus

Application

Anti-NMDAR2B Antibody is an antibody against NMDAR2B for use in IH, IP & WB.
Immunohistochemistry:
1:1,000-1:2,000 dilution of a previous lot was used on immunohistochemistry.

Immunoprecipitation:
3 μL of a previous lot of this antibody (under appropriate conditions) quantitatively immunoprecipitated all NMDAR2B in 200 μg of rat brain.

Western Blot Analysis:
1:100-1:2,000 dilution of a previous lot (NMDAR2B is present in high concentrations in the hippocampus).

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Target description

180 kDa

Physical form

Format: Purified
ImmunoAffinity Purified
Purified rabbit polyclonal lyophilized from ammonium bicarbonate (5 mM). Some residual salt may also be present. Reconstitute with 50 μL of PBS. Contains no preservatives.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Brain tissue, Rat Purkinje cell dendrites, rat hippocampus lysate, HEK 293 cells expressing NR2B.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subunit- and pathway-specific localization of NMDA receptors and scaffolding proteins at ganglion cell synapses in rat retina.
Zhang, J; Diamond, JS
The Journal of Neuroscience null
Proteome profiling of s-nitrosylated synaptosomal proteins by isobaric mass tags.
Teodora Stella Wijasa et al.
Journal of neuroscience methods, 291, 95-100 (2017-08-10)
Tzyy-Nan Huang et al.
Journal of psychiatry & neuroscience : JPN, 42(1), 37-47 (2017-02-25)
Human genetic studies have indicated that mutations in calcium/calmodulin-dependent serine protein kinase (CASK) result in X-linked mental retardation and autism-spectrum disorders. We aimed to establish a mouse model to study how Cask regulates mental ability. Because Cask encodes a multidomain
Josette S Lindahl et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 29(11), 2065-2073 (2004-05-13)
Phencyclidine (PCP) is a noncompetitive antagonist of the N-methyl-D-aspartate (NMDA) glutamate receptor subtype. It produces transient psychoses in normal individuals and exacerbates psychoses in schizophrenics. When administered to rodents, PCP elicits stereotypic behaviors including unrelenting head swaying, hyperlocomotion, and social
Distinct perisynaptic and synaptic localization of NMDA and AMPA receptors on ganglion cells in rat retina.
Zhang, J; Diamond, JS
The Journal of Comparative Neurology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service