Skip to Content
Merck
All Photos(1)

Key Documents

M-106

Supelco

Mycophenolic acid solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O6
CAS Number:
Molecular Weight:
320.34
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)CCC(O)=O

InChI

1S/C17H20O6/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3/h4,20H,5-8H2,1-3H3,(H,18,19)/b9-4+

InChI key

HPNSFSBZBAHARI-RUDMXATFSA-N

Gene Information

General description

Assayed in therapeutic drug monitoring to ensure patients remain within the drug′s therapeutic range, mycophenolic acid is an immunosuppressant drug and the active metabolite of the prodrug mycophenolate mofetil. This analytical standard is suitable for LC-MS/MS applications including therapeutic drug monitoring and other clinical or diagnostic applications.

Preparation Note

This product contains an ampule of 1 mL, at a concentration of 1.0 mg/mL in acetonitrile.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD25 blockade for refractory polymyositis.
Nicolò Pipitone et al.
Clinical and experimental rheumatology, 31(3), 474-474 (2013-03-08)
David Wofsy et al.
Arthritis and rheumatism, 65(6), 1586-1591 (2013-03-27)
Clinical trials of therapies for lupus nephritis have used many different primary outcome measures, ranging from complete response to time to end-stage renal disease. The objective of this study was to compare several possible outcome measures, using data from a
Mycophenolate mofetil in severe lupus nephritis: should post hoc analyses change treatment patterns?
Jonathan J Hogan et al.
American journal of kidney diseases : the official journal of the National Kidney Foundation, 61(5), 692-693 (2013-04-16)
Nathan R Selden et al.
Journal of neurosurgery. Pediatrics, 11(6), 643-652 (2013-04-16)
Infantile and late-infantile neuronal ceroid lipofuscinoses (NCLs) are invariably fatal lysosomal storage diseases associated with defects in lysosomal enzyme palmitoyl-protein thioesterase 1 (PPT-1) or tripeptidyl peptidase 1 (TPP1) activity. Previous preclinical studies have demonstrated that human CNS stem cells (HuCNS-SCs)
Yutaro Mori et al.
Nihon Jinzo Gakkai shi, 55(2), 177-184 (2013-05-02)
A 26-year-old man diagnosed with nephrotic syndrome was administered steroid monotherapy. Urinary protein excretion was 2-3 g/day despite the therapy. Percutaneous renal biopsy revealed Type I idiopathic membranoproliferative glomerulonephritis (IMPGN). Although intravenous steroid therapy at the dose of 1,000 mg/day

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service