Skip to Content
Merck
All Photos(4)

Key Documents

L5385

Sigma-Aldrich

α-Lactalbumin from bovine milk

Type I, ≥85% (PAGE), lyophilized powder

Synonym(s):

Bos d 4, Lactose synthase B protein, alpha-lactalbumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine milk

Quality Level

type

Type I

Assay

≥85% (PAGE)

form

lyophilized powder

concentration

≥85 % protein

technique(s)

cell culture | mammalian: suitable
electrophoresis: suitable

impurities

calcium, tested

solubility

H2O: soluble 10 mg/mL, clear to slightly hazy, colorless to faintly yellow

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

α-Lactalbumin is a small, globular, whey protein that has been found in all milk studied to date. It is a metalloprotein of approximately 14 kDa produced in the mammary glands.

Application

α-Lactalbumin from bovine milk has been used as a supplement of basal medium for various cell cultures. It has also been used as a marker for sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE).

Biochem/physiol Actions

α-Lactalbumin is the cheif protein in human milk. It consists of a single polypeptide chain with 8 cysteines which form disulfide bridges. α-Lactalbumin binds several metal ions, including calcium, which is thought to play a role in the regeneration of native α-lactalbumin from the reduced, denatured form. α-Lactalbumin also has a distinct zinc binding site that is thought to play a role in the binding of the lactose synthase complex. The mature protein consists of 123 amino acid residues (14 kD), and it has a three-dimensional structure with 1.7 Α° resolution, demonstrating four α-helices and a triple stranded antiparallel β-sheet.
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase.
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase. Site-directed mutagenesis of Asp87 or Asp88 to Ala completely abolishes the strong calcium binding affinity and reduces the stimulation of lactose synthase to <3.5% of the maximal rate.

Quality

Calcium saturated. May have traces of ammonium sulfate and sodium phosphate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Falk Bernsmann et al.
Journal of colloid and interface science, 344(1), 54-60 (2010-01-23)
We recently showed the possibility to build dopamine-melanin films of controlled thickness by successive immersions of a substrate in alkaline solutions of dopamine [F. Bernsmann, A. Ponche, C. Ringwald, J. Hemmerlé, J. Raya, B. Bechinger, J.-C. Voegel, P. Schaaf, V.
Daniel Rusu et al.
The Journal of nutrition, 140(2), 382-391 (2009-12-25)
Innate immunity depends on the efficiency of neutrophils to be activated rapidly to restore homeostasis. It can benefit from priming agents that enhance neutrophil capacity to respond more efficiently to a subsequent stimulation. Among natural products, a bovine whey protein
XiaoLu Geng et al.
Soft matter, 15(24), 4787-4796 (2019-05-08)
Formation of nanotubes from partially hydrolysed α-lactalbumin (α-La) was investigated at five pH values, two concentrations of α-La and two calcium levels. Nanotubes were formed under almost all combinations of the investigated factors, and for the first time the formation
Dmitri Nikitin et al.
Computational and structural biotechnology journal, 20, 1366-1377 (2022-04-08)
Cardio- and cerebrovascular diseases are leading causes of death and disability, resulting in one of the highest socio-economic burdens of any disease type. The discovery of bacterial and human plasminogen activators and their use as thrombolytic drugs have revolutionized treatment
Effect of different exogenous fatty acids on the cytosolic triacylglycerol content in bovine mammary cells
Vargas-Bello-Perez E, et al.
Animal Nutrition, 5(2), 202-208 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service