Skip to Content
Merck
All Photos(1)

Key Documents

T-049

Supelco

Tacrolimus solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H69NO12
CAS Number:
Molecular Weight:
804.02
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

CO[C@@H]1C[C@@H](CC[C@H]1O)\C=C(/C)[C@H]2OC(=O)[C@@H]3CCCCN3C(=O)C(=O)[C@]4(O)O[C@H]([C@H](C[C@@H](C)C\C(C)=C\[C@@H](CC=C)C(=O)C[C@H](O)[C@H]2C)OC)[C@H](C[C@H]4C)OC

InChI

1S/C44H69NO12/c1-10-13-31-19-25(2)18-26(3)20-37(54-8)40-38(55-9)22-28(5)44(52,57-40)41(49)42(50)45-17-12-11-14-32(45)43(51)56-39(29(6)34(47)24-35(31)48)27(4)21-30-15-16-33(46)36(23-30)53-7/h10,19,21,26,28-34,36-40,46-47,52H,1,11-18,20,22-24H2,2-9H3/b25-19+,27-21+/t26-,28+,29+,30-,31+,32-,33+,34-,36+,37-,38-,39+,40+,44+/m0/s1

InChI key

QJJXYPPXXYFBGM-LFZNUXCKSA-N

Gene Information

human ... FKBP1A(2280)

General description

Tacrolimus, also known as fujimycin, is an immunosuppressant used to prevent rejection of organ transplants. This Certified Spiking Solution® is suitable for use as starting material in the preparation of calibrators and controls for tacrolimus testing methods by LC-MS/MS. Tacrolimus is sold under trade names Progaf, Advagraf®, and Protopic®.

Legal Information

Advagraf is a registered trademark of Astellas Pharma Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Protopic is a registered trademark of Astellas Pharma Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Successful effect of tacrolimus in a 5-year-old boy presenting with repeated episodes of non-herpetic acute limbic encephalitis].
K Kishi et al.
No to hattatsu = Brain and development, 45(2), 152-154 (2013-05-09)
Tomohiro Tsuchiya et al.
Transplantation, 96(2), 198-204 (2013-06-25)
A prolonged-release formulation of tacrolimus (Tacrolimus QD) was developed to allow once-daily dosing and to have similar safety and efficacy profiles to twice-daily tacrolimus (Tacrolimus BID). This study compared the pharmacokinetics (PK) and renal pathology by protocol biopsy in de
Yingjing Wang et al.
Journal of biomedical nanotechnology, 9(2), 147-157 (2013-05-01)
As a traditional immunosuppressive drug, tacrolimus showed the potency in treating ulcerative colitis. In this study, a novel drug delivery vehicle achieved by self-assembly was applied to tacrolimus. During the preparation, amphiphilic copolymer MPEG-PCL was chosen to form the unique
Kensuke Kosugi et al.
Cranio : the journal of craniomandibular practice, 31(2), 140-150 (2013-06-26)
The authors performed a longitudinal study of the microstructural changes occurring in the mandibular condyle during osteoporosis using the findings obtained from micro-CT. The subjects used in this study were eight Sprague-Dawley rats. Among them, five were administered the immunosuppressant
Darla R Lower et al.
American journal of clinical pathology, 139(6), 788-792 (2013-05-22)
FK506 (tacrolimus), a macrolide immunosuppressant, is widely used in pediatric transplant patients, but a relatively narrow therapeutic window in children vs adults requires close and accurate monitoring of whole blood FK506 levels. High-pressure liquid chromatography/tandem mass spectrometry (HPLC/MS/MS)-based assays have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service