Skip to Content
Merck
All Photos(1)

Key Documents

E-006

Supelco

EDDP perchlorate

ampule of 10 mg, certified reference material, Cerilliant®

Synonym(s):

EDDP perchlorate, 2-Ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N · HClO4
CAS Number:
Molecular Weight:
377.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

solid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 10 mg

manufacturer/tradename

Cerilliant®

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

neat

storage temp.

−20°C

General description

A Certified Reference Material of the major urinary metabolite of methadone. Methadone, sold under various trade names such as Symoron, Amidone or Methadose®, is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. Suitable uses for this reference standard include as starting material in preparation of calibrators and controls for LC/MS or GC/MS methods in methadone testing for applications in clinical toxicology or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Methadose is a registered trademark of Mallinckrodt Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cristiana Gambelunghe et al.
Annals of clinical and laboratory science, 43(1), 22-30 (2013-03-07)
It may be advantageous to use sweat, rather than blood or urine, to monitor individuals' drug exposure for the purposes of drug treatment programs, employment initiatives, and forensic investigations. Forty-eight patients receiving methadone at the Public Service for the Treatment
Ricarda Jantos et al.
Forensic science international, 226(1-3), 254-260 (2013-02-21)
Body fluids and tissues in 16 methadone (MTD)-associated fatalities were investigated to find out whether analysis of MTD and its metabolite 2-ethylidine-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP) following enantioselective separation of both compounds may assist in the interpretation of MTD findings. Individual case histories
Karen Marie Dollerup Holm et al.
Journal of analytical toxicology, 36(7), 487-496 (2012-07-11)
We developed a method based on liquid chromatography coupled with tandem mass spectrometry to quantify individual enantiomers of methadone and its primary metabolite, R/S-2-ethyl-1,5-dimethyl-3,3-diphenylpyrrolinium (EDDP), in postmortem blood and brain tissue. Samples were prepared with a Tecan Evo robotic system.
Maksymilian Kulza et al.
Przeglad lekarski, 67(10), 925-928 (2010-01-01)
Methadone maintenance therapy is one of the most popular treatments for opiate addiction. The easiest and the most reliable way to monitor the accuracy of the therapy is determination of methadone and its metabolites in biological material. The aim of
Mathew George et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 8(4), 432-435 (2012-08-18)
Reports describing methadone overdose in adult and pediatric patient populations are not uncommon. Reports in the preterm neonate patient population, however, are infrequent even though the utilization of methadone in that population continues to increase. Significant age-related pharmacokinetic differences complicate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service