Skip to Content
Merck
All Photos(5)

Key Documents

G0251

Sigma-Aldrich

β-Glucuronidase from bovine liver

Type B-1, ≥1,000,000 units/g solid

Synonym(s):

Bovine Liver beta-Glucuronidase, β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type B-1

form

solid

specific activity

≥1,000,000 units/g solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Glucuronidase (GUSB) is a lysosomal enzyme, found in animals, plants and bacteria. This exoglycosidase shows high activity in spleen, liver and kidney. Bovine liver β-glucuronidase (BLG) is homotetramer with a molecular weight of 290 kDa.

Application

β-Glucuronidase from bovine liver has been used in β-glucuronidase inhibition assay to study the β-glucuronidase inhibitory effect of extracts and isolated actives from Phyllanthus amarus. It has also been used to perform deglucuronidation.
β-glucuronidase is used as a reporter gene in GUS assays to monitor gene expression.

Biochem/physiol Actions

β-Glucuronidase (GUSB) deficiency can result in mucopolysaccharidosis type VII (MPSVII), which leads to store lysosome in the brain. Integration of β-glucuronides of steroid hormones into tissues can be done with the help of GUSB.
β-glucuronidase catalyzes the breakdown of complex carbohydrates. In humans it converts conjugated bilirubin into the unconjugated form, making bilirubin suitable for reabsorption.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 5.0 (30 min assay).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Jakobsen et al.
The Journal of pharmacy and pharmacology, 33(2), 89-92 (1981-02-01)
Sulphinpyrazone is an antiplatelet in vivo and in vitro. Two active metabolites, a sulphide (S) and a hydroxylated sulphide (S-OH) have been identified in rabbit and human plasma and a selective and sensitive g.c.-m.s.-method for quantitative determination of the sulphide
A D Fraser
Journal of analytical toxicology, 11(6), 263-266 (1987-11-01)
Alprazolam (Xanax) and triazolam (Halcion) are relatively new triazolobenzodiazepines that are anxiolytic and hypnotic. This study assesses the reactivity of these drugs and their major metabolites in the EMIT d.a.u. benzodiazepine metabolite assay. Analytical standards of drugs and metabolites and
Determination of furosemide with its acyl glucuronide in human plasma and urine by means of direct gradient high-performance liquid chromatographic analysis with fluorescence detection Preliminary pharmacokinetics and effect of probenecid
Vree TB, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 655(1), 53-62 (1994)
Chloé Hugbart et al.
Toxicology in vitro : an international journal published in association with BIBRA, 66, 104842-104842 (2020-04-14)
Ethanol is the most frequently psychoactive substance used in the world, leading to major public health problems with several millions of deaths attributed to alcohol consumption each year. Metabolism of ethanol occurs mainly in the liver via the predominant oxidative
Uzma Salar et al.
European journal of medicinal chemistry, 125, 1289-1299 (2016-11-26)
Current study is based on the biology-oriented drug synthesis (BIODS) of 2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethyl aryl carboxylate derivatives 1-26, by treating metronidazole with different aryl and hetero-aryl carboxylic acids in the presence of 1,1'-carbonyl diimidazole (CDI) as a coupling agent. Structures of all

Protocols

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service