Skip to Content
Merck
All Photos(1)

Key Documents

E-022

Supelco

EDDP perchlorate solution

1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Methadone metabolite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClNO4
CAS Number:
Molecular Weight:
377.86
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as pyrolinium)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

C[N+]1=C(CC)C(C2=CC=CC=C2)(C3=CC=CC=C3)CC1C.O=Cl(=O)([O-])=O

InChI

1S/C20H24N.ClHO4/c1-4-19-20(15-16(2)21(19)3,17-11-7-5-8-12-17)18-13-9-6-10-14-18;2-1(3,4)5/h5-14,16H,4,15H2,1-3H3;(H,2,3,4,5)/q+1;/p-1

InChI key

FICCBMGBBWWZRF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike® solution of the major urinary metabolite of methadone. Methadone, sold under various trade names such as Symoron, Amidone or Methadose®, is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. Suitable uses for this reference standard include as starting material in preparation of calibrators and controls for LC/MS or GC/MS methods in methadone testing for applications in clinical toxicology or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Methadose is a registered trademark of Mallinckrodt Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rosa Boleda et al.
Chemosphere, 84(11), 1601-1607 (2011-06-15)
A total of seventy samples of drinking water were tested for non-controlled and illicit drugs. Of these, 43 were from Spanish cities, 15 from seven other European countries, three from Japan and nine from seven different Latin American countries. The
Ana de Castro et al.
Therapeutic drug monitoring, 33(4), 443-452 (2011-07-12)
The purpose was to explore methadone and 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP) umbilical cord disposition, correlate with maternal methadone dose and neonatal outcomes, and evaluate the window of drug detection in umbilical cord of in utero illicit drug exposure. Subjects comprised 19 opioid-dependent
Edijs Leimanis et al.
Journal of analytical toxicology, 36(4), 239-249 (2012-04-19)
Methadone is used to treat moderate to severe pain in patients not responsive to non-narcotic analgesics and for maintenance treatment of opioid addiction. Methadone is primarily metabolized by N-demethylation to an inactive metabolite 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidene (EDDP) by CYP3A4 and CYP2B6. Establishing
Ana de Castro et al.
Clinical chemistry, 57(3), 449-458 (2011-01-20)
Few investigations have used placenta as an alternative matrix to detect in utero drug exposure, despite its availability at the time of birth and the large amount of sample. Methadone-maintained opioid-dependent pregnant women provide a unique opportunity to examine the
Wenjie Jessie Lu et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(8), 1308-1313 (2010-04-23)
The peripheral conversion of testosterone to estradiol by aromatase is the primary source of endogenous estrogen in postmenopausal women. Studies indicating that placental aromatase is able to metabolize methadone to its primary metabolite, 2-ethylidene-1, 5-dimethyl-3, 3-diphenylpyrrolidin (EDDP), led us to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service