Skip to Content
Merck
All Photos(1)

Documents

D-048

Supelco

N-Desmethylclozapine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClN4
CAS Number:
Molecular Weight:
312.80
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCNCC4

InChI

1S/C17H17ClN4/c18-12-5-6-15-16(11-12)21-17(22-9-7-19-8-10-22)13-3-1-2-4-14(13)20-15/h1-6,11,19-20H,7-10H2

InChI key

JNNOSTQEZICQQP-UHFFFAOYSA-N

General description

N-Desmethylclozapine is an active metabolite of Clozapine, an atypical antipsychotic drug used for treatment of schizophrenia. Clozapine is sold under the trade names Clozaril® and FrazaClo in the US. This Certified Snap-N-Spike® Solution is suitable for use in urine drug testing, clinical toxicology, forensic analysis, and clinical monitoring by LC-MS/MS or GC/MS.

Application



  • Pharmacokinetic analysis of clozapine and N-Desmethylclozapine: A study conducted by Mach et al. (2024) analyzed clinical and pharmacological factors that influence serum levels of clozapine and its metabolite N-Desmethylclozapine, offering insights into dosing strategies and metabolic pathways relevant to schizophrenia treatment (Mach et al., 2024).


  • Chromatographic profiling for clozapine and its metabolites: Hrichi et al. (2024) reviewed chromatographic methods for analyzing antipsychotic drugs, including clozapine and N-Desmethylclozapine, to improve the accuracy of pharmacokinetic studies and enhance drug monitoring capabilities (Hrichi et al., 2024).


  • Influence of lifestyle factors on clozapine metabolism: Research by Flanagan et al. (2024) explored the impact of coffee and chocolate ingestion on the plasma concentrations of clozapine and its metabolite N-Desmethylclozapine, providing data crucial for understanding how dietary habits affect antipsychotic drug levels (Flanagan et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Clozaril is a registered trademark of Sandoz, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jalagam Anjaneyulu et al.
Journal of Ayurveda and integrative medicine, 11(4), 440-447 (2020-09-27)
Globally, there is increased incidence of Parkinson's Disease (PD), which is the second most common age-related neurodegenerative disease. The currently available PD-therapeutics provide only symptomatic relief. Thus, there is an urgent need to devise an effective and safe treatment strategy
Maria C Olianas et al.
European journal of pharmacology, 660(2-3), 341-350 (2011-04-26)
We have previously reported that N-desmethylclozapine (NDMC), a major clozapine metabolite, acts as a δ-opioid receptor agonist. Here, we show that in different cellular systems NDMC regulates protein kinase B/Akt (Akt) signaling through the activation of δ-opioid receptors. In Chinese
K Koda et al.
British journal of pharmacology, 162(3), 763-772 (2010-10-21)
We have previously reported that galantamine, a weak acetylcholinesterase inhibitor, improves prepulse inhibition (PPI) deficits in mice reared in social isolation. ACh receptors are involved in the underlying mechanism of PPI, but whether rearing in social isolation causes dysfunction of
Kathryn K Erickson-Ridout et al.
Pharmacogenetics and genomics, 22(8), 561-576 (2012-05-09)
Clozapine (CLZ) is an FDA approved second-generation antipsychotic for refractory schizophrenia, and glucuronidation is an important pathway in its metabolism. The aim of this study was to fully characterize the CLZ glucuronidation pathway and examine whether polymorphisms in active glucuronidating
Yousheng Cai et al.
Scientific reports, 10(1), 10444-10444 (2020-06-28)
Parkinson's disease (PD) is a common dyskinesia disease, the mitochondrial unfolded protein response (mtUPR) may be directly or indirectly involved in the occurrence and development of PD, although the exact mechanism is unclear. We established a dopaminergic neuronal-like cell model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service