Skip to Content
Merck
All Photos(1)

Key Documents

O-024

Supelco

Olanzapine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4S
CAS Number:
Molecular Weight:
312.43
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24

InChI

1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3

InChI key

KVWDHTXUZHCGIO-UHFFFAOYSA-N

General description

An analytical reference standard for use in LC/MS or GC/MS applications including clinical toxicology analysis or therapeutic drug monitoring. Marketed under the trade name Zyprexa®, olanzapine is an atypical antipsychotic drug used in the treatment of schizophrenia and bipolar disorder.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zyprexa is a registered trademark of Eli Lilly and Co.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean A Milstein et al.
PloS one, 8(2), e57308-e57308 (2013-02-26)
Antipsychotic drugs are increasingly used in children and adolescents to treat a variety of psychiatric disorders. However, little is known about the long-term effects of early life antipsychotic drug treatment. Most antipsychotic drugs are potent antagonists or partial agonists of
Heating under high-frequency inductive conditions: application to the continuous synthesis of the neurolepticum olanzapine (Zyprexa).
Jan Hartwig et al.
Angewandte Chemie (International ed. in English), 52(37), 9813-9817 (2013-07-31)
Robin H Schmidt et al.
The Journal of pharmacology and experimental therapeutics, 347(1), 126-135 (2013-08-09)
Olanzapine (OLZ), an effective treatment of schizophrenia and other disorders, causes weight gain and metabolic syndrome. Most studies to date have focused on the potential effects of OLZ on the central nervous system's mediation of weight; however, peripheral changes in
Chantal Henry et al.
BMC psychiatry, 13, 79-79 (2013-03-21)
One of the major issues in clinical practice is the accurate differential diagnosis between mixed states and depression, often leading to inappropriate prescriptions of antidepressants in mixed states, and as a consequence, increasing the risk of manic switch and suicide.
A case of atypical antipsychotic-induced somnambulism: a class effect.
Yael Dagan et al.
The Journal of clinical psychiatry, 74(4), 370-370 (2013-05-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service