Skip to Content
Merck
All Photos(1)

Documents

H-030

Supelco

Haloperidol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClFNO2
CAS Number:
Molecular Weight:
375.86
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3

InChI

1S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2

InChI key

LNEPOXFFQSENCJ-UHFFFAOYSA-N

Gene Information

human ... HTR2A(3356)

General description

Haloperidol, sold under the trade name Haldol®, is a typical antipsychotic which functions as a dopamine inverse agonist. This Certified Snap-N-Spike® Solution is suitable for use in LC/MS or GC/MS applications such as clinical toxicology, urine drug testing, and forensic analysis. This butyrophenone analog is used to treat schizophrenia and delirium.

Application


  • Haloperidol solution for schizophrenia treatment: Utilized primarily as an antipsychotic medication, haloperidol is effective in managing symptoms of schizophrenia by acting as a dopamine receptor antagonist, offering a solution for reducing psychotic episodes and improving patient outcomes (Heindorf and Keller, 2024).

  • Haloperidol solution mechanism of action: As a neuroleptic drug, haloperidol operates by blocking dopamine receptors in the brain, specifically in areas associated with mood, behavior, and cognition, which is crucial for its effectiveness in treating disorders like schizophrenia and bipolar disorder (Liu et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Haldol is a registered trademark of Johnson & Johnson
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dan Shan et al.
Schizophrenia research, 144(1-3), 1-8 (2013-01-30)
Glutamate transporters facilitate the buffering, clearance and cycling of glutamate and play an important role in maintaining synaptic and extrasynaptic glutamate levels. Alterations in glutamate transporter expression may lead to abnormal glutamate neurotransmission contributing to the pathophysiology of schizophrenia. In
Norma Adán et al.
The Journal of clinical investigation, 123(9), 3902-3913 (2013-08-03)
Chondrocytes are the only cells in cartilage, and their death by apoptosis contributes to cartilage loss in inflammatory joint diseases, such as rheumatoid arthritis (RA). A putative therapeutic intervention for RA is the inhibition of apoptosis-mediated cartilage degradation. The hormone
Gerard W K Hugenholtz et al.
The Journal of clinical psychiatry, 67(6), 897-903 (2006-07-20)
To determine the doses of haloperidol as a comparator drug in randomized controlled trials (RCTs) with atypical antipsychotics in patients with schizophrenia and to compare these doses with the officially recommended doses for haloperidol in the United States and the
Paul Perkins et al.
The Cochrane database of systematic reviews, (2)(2), CD006271-CD006271 (2009-04-17)
Nausea and vomiting are common symptoms of patients with terminal, incurable illnesses and can be distressing. The primary objective of the review was to evaluate the efficacy and adverse events associated with the use of haloperidol for the treatment of
Marianne Klemp et al.
Journal of clinical psychopharmacology, 31(6), 698-704 (2011-10-25)
The objective of the study was to examine the efficacy and the degree of adverse effects connected with atypical neuroleptic drugs and haloperidol by using a previously described Bayesian statistical method that includes both direct and indirect comparisons simultaneously. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service