Skip to Content
Merck
All Photos(1)

Key Documents

LM4000

Avanti

19:0 Cholesterol ester

Avanti Research - A Croda Brand LM4000, (methanol/chloroform/toluene (50:25:25) solution)

Synonym(s):

cholest-5-en-3β-yl nonadecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H82O2
CAS Number:
Molecular Weight:
667.14
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

(methanol/chloroform/toluene (50:25:25) solution)

packaging

pkg of 1 × 1 mL (LM4000-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM4000

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](OC(CCCCCCCCCCCCCCCCCC)=O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC(C)C

InChI key

VHYWECIJXTVRSG-TVDLSCFRSA-N

General description

19:0 Cholesterol ester is the central core of lipoprotein particles. CE formation is catalyzed by the enzyme lecithin cholesterol acyl transferase (LCAT).[1]

Application

19:0 Cholesterol ester has been used as a quantitative lipid standard to monitor different lipid extract concentrations.[2]

Biochem/physiol Actions

CE hydrolysis mediates reverse cholesterol transport in macrophage foam cells.[3]

Packaging

2 mL Amber Glass Sealed Ampule (LM4000-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes, Liver,Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The critical role of neutral cholesterol ester hydrolase 1 in cholesterol removal from human macrophages.
Igarashi M, et al.
Circulation Research, 107(11), 1387-1395 (2010)
Mammalian acyl-CoA:cholesterol acyltransferases
Buhman KF, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1529(1), 142-154 (2000)
A comprehensive method for lipid profiling by liquid chromatography-ion cyclotron resonance mass spectrometry
Fauland A, et al.
Journal of Lipid Research, 52(12), 2314-2322 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service