Skip to Content
Merck
All Photos(1)

Key Documents

330709W

Avanti

SPLASH II LIPIDOMIX Mass Spec Standard

Avanti Research - A Croda Brand

Synonym(s):

SPLASH II LIPIDOMIX Mass Spec Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

form

methanol solution

packaging

pkg of 1 × 1 mL (330709W-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL amber glass sealed ampule

Components

The SPLASH II LIPIDOMIX Mass Spec Standard is supplied in 1 mL of methanol and contains the following components:

Mixture ComponentTarget Conc. (μM)
  • 15:0-18:1 (d7) PC 210
  • 15:0-18:1 (d7) PE7 7
  • 15:0-18:1 (d7) PS (Na salt) 10
  • 15:0-18:1 (d7) PI (NH4 salt) 10
  • 18:1-(d7) Lyso PC 45
  • 18:1-(d7) Lyso PE 1
  • 18:1 (d7) cholesterol ester 530
  • C18(plasm)-18:1(d9) PC 10
  • 15:0-18:1-(d7) DAG 20
  • 15:0-18:1-(d7):15:0 TAG 70
  • d18:1-18:1-(d9) SM 40
  • C18(plasm)-18:1(d9) PE 0.1

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 160μg/mL

  • 791638C15:0-18:1-d7-PE, Avanti Research - A Croda Brand 5μg/mLSDS

  • 791639C15:0-18:1-d7-PS, Avanti Research - A Croda Brand 8μg/mLSDS

  • 791641C15:0-18:1-d7-PI, Avanti Research - A Croda Brand 8μg/mLSDS

  • 791643C18:1-d7 Lyso PC, Avanti Research - A Croda Brand 25μg/mLSDS

  • 791644C18:1-d7 Lyso PE, Avanti Research - A Croda Brand 791644C 0.5μg/mLSDS

  • 700185M18:1 Chol (D7) ester, 25,26,26,26,27,27,27-heptadeuteriocholest-5-en-3β-ol (9Z-octadecenoate), methylene chloride solution 350μg/mLSDS

  • 852475CC18(plasm)-18:1(d9) PC, Avanti Research - A Croda Brand, chloroform solution 8μg/mLSDS

  • 791647C15:0-18:1-d7 DG, Avanti Research - A Croda Brand 791647C 12μg/mLSDS

  • 791648C15:0-18:1-d7-15:0 TG, Avanti Research - A Croda Brand 791648C 55μg/mLSDS

  • 791649C18:1-d9 SM, Avanti Research - A Croda Brand 30μg/mLSDS

  • 852474CC18(plasm)-18:1(d9) PE, Avanti Research - A Croda Brand, chloroform solution 0.07μg/mLSDS

See All (12)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Triebl et al.
Journal of lipid research, 61(1), 105-115 (2019-11-17)
Quantitative MS of human plasma lipids is a promising technology for translation into clinical applications. Current MS-based lipidomic methods rely on either direct infusion (DI) or chromatographic lipid separation methods (including reversed phase and hydrophilic interaction LC). However, the use
Maximilian Schütter et al.
Cell, 180(1), 135-149 (2019-12-31)
Autophagy is a conserved catabolic homeostasis process central for cellular and organismal health. During autophagy, small single-membrane phagophores rapidly expand into large double-membrane autophagosomes to encapsulate diverse cargoes for degradation. It is thought that autophagic membranes are mainly derived from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service