Skip to Content
Merck
All Photos(1)

Key Documents

880130C

Avanti

18:1 PEG2000 PE

Avanti Research - A Croda Brand 880130C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (880130C-25mg)
pkg of 2 × 20 mL (880130C-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 880130C

concentration

10 mg/mL (880130C-25mg)
25 mg/mL (880130C-1g)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C133H260NO55P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-131(135)186-128-130(189-132(136)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)129-188-190(138,139)187-37-36-134-133(137)185-127-126-184-125-124-183-123-122-182-121-120-181-119-118-180-1

InChI key

QXTYPWABYAAAPJ-FCKVYVLXSA-N

Application

18:1 PEG2000 PE has been used as a component of quantum dot (QD) solution. It has also been used in fusion assay to study the inter-bilayer and bilayer-substrate interactions.

Biochem/physiol Actions

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine is a neutral lipid. It serves as a helper lipid for cationic liposomes. This product might be used for PEGylation (polyethylene glycol) of nanoparticles to increase in vivo stability. It also promotes efficient delivery.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (880130C-1g)
5 mL Clear Glass Sealed Ampule (880130C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of cationic liposomes for delivery of nucleic acids
Shim G, et al.
Asian Journal of Pharmaceutical Sciences , 8(2), 72-80 (2013)
Wide-field three-photon excitation in biological samples
Rowlands CJ, et al.
Light: Science & Applications, 6(5), e16255-e16255 (2017)
Fusion of single proteoliposomes with planar, cushioned bilayers in microfluidic flow cells
Karatekin E and Rothman JE
Nature Protocols, 7, 903-903 (2012)
Carol Cho et al.
Nature communications, 10(1), 5764-5764 (2019-12-19)
The fundamental unit of chromatin, the nucleosome, is an intricate structure that requires histone chaperones for assembly. ATAD2 AAA+ ATPases are a family of histone chaperones that regulate nucleosome density and chromatin dynamics. Here, we demonstrate that the fission yeast ATAD2
Mudassar Mumtaz Virk et al.
Langmuir : the ACS journal of surfaces and colloids, 34(1), 395-405 (2017-12-13)
Hybrid vesicles, comprising blends of amphiphilic block copolymers and phospholipids, have attracted significant attention recently because of their unique combination of chemical and physical properties. We report a method to make unilamellar hybrid vesicles with diameters of 100 nm by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service