Skip to Content
Merck
All Photos(1)

Key Documents

860901P

Avanti

15:0-18:1-15:0 D5 TG

Avanti Research - A Croda Brand 860901P, powder

Synonym(s):

1,3(d5)-dipentadecanoyl-2-(9Z-octadecenoyl)-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H91D5O6
CAS Number:
Molecular Weight:
810.34
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860901P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860901P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C(C([2H])([2H])OC(CCCCCCCCCCCCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)C([2H])([2H])OC(CCCCCCCCCCCCCC)=O

InChI

1S/C51H96O6/c1-4-7-10-13-16-19-22-25-26-27-30-33-36-39-42-45-51(54)57-48(46-55-49(52)43-40-37-34-31-28-23-20-17-14-11-8-5-2)47-56-50(53)44-41-38-35-32-29-24-21-18-15-12-9-6-3/h25-26,48H,4-24,27-47H2,1-3H3/b26-25-/i46D2,47D2,48D

InChI key

YUNYDLOKHYJQAT-OFSSTFKFSA-N

General description

15:0-18:1-15:0 D5 TG or 1,3(d5)-dipentadecanoyl-2-(9Z-octadecenoyl)-glycerol is a prostaglandin deuterated isotope.

Application

15:0-18:1-15:0 D5 TG has been used as an internal standard to measure the triglycerol in the experimental sample by ultra performance liquid chromatography - tandem mass spectrometer (UPLC-MS/MS) and high-performance liquid chromatography (HPLC)-MS/MS.

Packaging

5 mL Clear Glass Sealed Ampule (860901P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zongyuan Wu et al.
Food & function, 9(10), 5103-5114 (2018-09-13)
Atherosclerosis is a chronic disease associated with oxidative stress and inflammatory activation and is the main underlying trigger for cardiovascular disease. There are many cardiovascular health products in the market; in order to evaluate the effect of these products, in
Qin-Li Wan et al.
Oxidative medicine and cellular longevity, 2019, 5768953-5768953 (2019-06-30)
Aging is a complex life process, and a unified view is that metabolism plays key roles in all biological processes. Here, we determined the lipidomic profile of Caenorhabditis elegans (C. elegans) using ultraperformance liquid chromatography high-resolution mass spectrometry (UPLC-HRMS). Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service