Skip to Content
Merck
All Photos(2)

Key Documents

850706P

Avanti

16:1 PE

1,2-dipalmitoleoyl-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

1,2-di-(9Z-hexadecenoyl)-sn-glycero-3-phosphoethanolamine; PE(16:1(9Z)/16:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H70NO8P
CAS Number:
Molecular Weight:
687.93
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (850706P-200mg)
pkg of 1 × 25 mg (850706P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850706P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCC)=O)COC(CCCCCCC/C=C\CCCCCC)=O

InChI

1S/C37H70NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h13-16,35H,3-12,17-34,38H2,1-2H3,(H,41,42)/b15-13-,16-14-/t35-/m1/s1

InChI key

PGPMCWZMPPZJML-NAFNZUQFSA-N

General description

Phosphatidylethanolamine is an aminophospholipid, which is found in eukaryotic and prokaryotic cells.[1] It is one of the most abundant glycerophospholipids in biological membranes.[2] Phosphatidylethanolamine is found in nervous tissue.[3]

Application

16:1 PE has been used to assess the linearity of response of lipid classes.[4] It has also been used as an internal standard for the determination of plasmenyl-PC and plasmenyl-PE of human heart mitochondria using a Bligh -Dyer extraction method.[5]

Biochem/physiol Actions

Phosphatidylethanolamine forms pure lipid structures.[6] It maintains membrane fluidity in eukaryotic cells.[7] Phosphatidylethanolamine plays a critical role in autophagy, cell division and protein folding. It acts as a precursor for the synthesis of various protein modifications. Phosphatidylethanolamine functions as an intermediate for the synthesis of glycerophospholipid classes.[2]

Packaging

5 mL Clear Glass Sealed Ampule (850706P-200mg)
5 mL Clear Glass Sealed Ampule (850706P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
An Introduction to Biological Membranes: Composition, Structure and Function (2016)
Cytochrome c is an oxidative stress-activated plasmalogenase that cleaves plasmenylcholine and plasmenylethanolamine at the sn-1 vinyl ether linkage
Jenkins CM, et al.
The Journal of Biological Chemistry, 293(22), 8693-8709 (2018)
High-Throughput Plasma Lipidomics: Detailed Mapping of the Associations with Cardiometabolic Risk Factors
Huynh K, et al.
Cell Chemical Biology, 26(1), 71-84 (2019)
Phosphatidylethanolamine and phosphatidylcholine biosynthesis by the Kennedy pathway occurs at different sites in Trypanosoma brucei
Farine L, et al.
Scientific reports, 5, 16787-16787 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service