Skip to Content
Merck
All Photos(1)

Documents

W506907

Sigma-Aldrich

(1S)-(−)-Verbenone

≥93%

Synonym(s):

(1S,5S)-2-Pinen-4-one, (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907623
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥93%

optical activity

[α]20/D −140°, c = 10 in ethanol

refractive index

n20/D 1.496 (lit.)

bp

227-228 °C (lit.)

density

0.975 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

camphoraceous; minty; spicy

SMILES string

CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

InChI

1S/C10H14O/c1-6-4-9(11)8-5-7(6)10(8,2)3/h4,7-8H,5H2,1-3H3/t7-,8+/m0/s1

InChI key

DCSCXTJOXBUFGB-JGVFFNPUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Anti-Inflammatory and Antinociceptive Activities of the Essential Oil of Tagetes parryi A. Gray (Asteraceae) and Verbenone.: This article reports on the anti-inflammatory and pain-relief effects of (1S)-(−)-Verbenone as part of the essential oil of Tagetes parryi, suggesting potential therapeutic applications for pain management and inflammation (González-Velasco et al., 2022).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mitsuo Miyazawa et al.
Drug metabolism and disposition: the biological fate of chemicals, 31(8), 1049-1053 (2003-07-18)
(-)-Verbenone, a monoterpene bicyclic ketone, is a component of the essential oil from rosemary species such as Rosmarinus officinalis L., Verbena triphylla, and Eucalyptus globulus and is used for an herb tea, a spice, and a perfume. In this study
R A Progar et al.
Journal of economic entomology, 106(1), 221-228 (2013-03-02)
Mountain pine beetle, Dendroctonus ponderosae Hopkins (Coleoptera: Curculionidae: Scolytinae), is among the primary causes of mature lodgepole pine, Pinus contorta variety latifolia mortality. Verbenone is the only antiaggregant semiochemical commercially available for reducing mountain pine beetle infestation of lodgepole pine.
Mitsuo Miyazawa et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 793(2), 291-296 (2003-08-09)
A sensitive assay for the determination of (-)-verbenone 10-hydroxylation catalyzed by rat liver microsomes was developed using high-performance liquid chromatography. Verbenone was incubated in vitro with liver microsomes of untreated rats and rats treated with phenobarbital and the products thus
Christopher J Fettig et al.
Journal of economic entomology, 105(5), 1668-1680 (2012-11-20)
The western pine beetle, Dendroctonus brevicomis LeConte (Coleoptera: Curculionidae, Scolytinae), is a major cause of ponderosa pine, Pinus ponderosa Douglas ex Lawson, mortality in much of western North America. We review several years of research that led to the identification
Michael Pescheck et al.
Journal of industrial microbiology & biotechnology, 36(6), 827-836 (2009-03-27)
A closed gas loop bioprocess was developed to improve fungal biotransformation of monoterpenes. By circulating monoterpene-saturated process gas, the evaporative loss of the volatile precursor from the medium during the biotransformation was avoided. Penicillium solitum, isolated from kiwi, turned out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service