Skip to Content
Merck
All Photos(3)

Documents

N32407

Sigma-Aldrich

Bicyclo[2.2.1]hept-2-ene

99%

Synonym(s):

2-Norbornene, Norbornylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10
CAS Number:
Molecular Weight:
94.15
Beilstein:
1098544
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

96 °C (lit.)

mp

44-46 °C (lit.)

SMILES string

C1C[C@H]2C[C@@H]1C=C2

InChI

1S/C7H10/c1-2-7-4-3-6(1)5-7/h1-2,6-7H,3-5H2/t6-,7+

InChI key

JFNLZVQOOSMTJK-KNVOCYPGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bicyclo[2.2.1]hept-2-ene is a cyclic olefin used in the transition metal-catalyzed dimerization of alkene.

The vinylic polymerization of bicyclo[2.2.1]hept-2-ene (norbornene) with Co(II) compounds and the metallocene [η5-(C5Me5)Co-η2-Cl]2, in chlorobenzene activated with methylaluminoxane (MAO) has been reported. Co(II) catalyzed polymerization of bicyclo[2.2.1]hept-2-ene in the presence of ethene has been reported.

Application

Bicyclo[2.2.1]hept-2-ene may be used in the preparation of cis,exo-2,3-diarylsubstituted bicyclo[2.2.1]heptanes or bicyclo [2.2.1] hept-2-enes.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1 - Repr. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Co (II)-catalyzed polymerization of bicyclo [2.2. 1] hept-2-ene in the presence of ethene.
Alt FP and Heitz W.
Acta Polym., 49(9), 477-481 (1998)
A new palladium catalyzed synthesis of cis, exo-2, 3-diarylsubstituted bicyclo [2.2. 1] heptanes or bicyclo [2.2. 1] hept-2-enes.
Catellani M, et al.
Tetrahedron, 45(16), 5263-5268 (1989)
Srivari Chandrasekhar et al.
Chemical communications (Cambridge, England), (14)(14), 1548-1550 (2006-04-01)
The oligomers of constrained cis-exo-beta-norbornene amino acid were synthesised and characterised by extensive NMR, CD, IR and MD studies. The results showed the formation of both right and left handed consecutive 6-membered hydrogen-bonded strands for [2S,3R] and [2R,3S] enantiomers, respectively.
Carolina Blanco et al.
Dalton transactions (Cambridge, England : 2003), 41(23), 6980-6991 (2012-04-27)
Palladium complexes bearing monodentate and bidentate phosphine ligands (1-7) were synthesised and used as catalyst precursors in the methoxycarbonylation of norbornene. The catalytic systems bearing ligands 1, 3 and 4 afforded excellent conversions (>99%) and selectivity of the ester (>99%).
Vinylic polymerization of bicyclo [2.2. 1] hept-2-ene by Co (II)-catalysis.
Alt FP and Heitz W.
Macromolecular Chemistry and Physics, 199(9), 1951-1956 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service