Skip to Content
Merck
All Photos(3)

Key Documents

W205532

Sigma-Aldrich

Isoamyl acetate

greener alternative

natural, ≥97%, FCC, FG

Synonym(s):

Isopentyl acetate, Acetic acid 3-methylbutyl ester, Isoamyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2055
Beilstein:
1744750
EC Number:
Council of Europe no.:
214
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.024
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

4.5 (vs air)

vapor pressure

5 mmHg ( 25 °C)

Assay

≥97%

form

liquid

autoignition temp.

680 °F

expl. lim.

7.5 %

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.4 (lit.)

bp

142 °C/756 mmHg (lit.)

mp

−78 °C (lit.)

solubility

ethanol: soluble 1ml/3ml, clear, colorless (60%ethanol)

density

0.876 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

banana; fruity; sweet

SMILES string

CC(C)CCOC(C)=O

InChI

1S/C7H14O2/c1-6(2)4-5-9-7(3)8/h6H,4-5H2,1-3H3

InChI key

MLFHJEHSLIIPHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoamyl acetate is the major ginjo-flavor components of Japanese sake (Ginjo-shu). It is effective in inactivating various microorganisms. Synthesis of isoamyl acetate by lipase-catalyzed transesterification of ethyl acetate in n-hexane has been reported. It is flavor-active compound present in beer.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Impact of non-Saccharomyces yeasts derived from traditional fermented foods on beer aroma: Analysis based on HS-SPME-GC/MS combined with chemometrics.: This study investigates the influence of non-Saccharomyces yeasts on beer aroma using headspace solid-phase microextraction-gas chromatography/mass spectrometry (HS-SPME-GC/MS) and chemometric analysis. Isoamyl acetate was identified as a significant compound contributing to the aroma profile, enhancing fruity and sweet notes in the beer (Cao et al., 2024).

  • Investigating the impact of various sorghum types on the key aroma compounds of Sichuan Xiaoqu Baijiu through application of the sensomics approach.: This research explores the effect of different sorghum varieties on the aroma compounds of Sichuan Xiaoqu Baijiu, a Chinese liquor. Isoamyl acetate was highlighted as a key volatile compound contributing to the characteristic fruity aroma of Baijiu (Ma et al., 2024).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rizzi et al.
Enzyme and microbial technology, 14(9), 709-714 (1992-09-01)
Isoamyl acetate was synthesized by lipase-catalyzed transesterification of ethyl acetate in n-hexane. The selectivity and rates of ester formation decreased when water content of the immobilized enzyme exceeded 3% (w/w). Experimental observations clearly indicate that the substrates as well as
Carrier-free, continuous primary beer fermentation.
Pires EJ, et al.
Journal of the Institute of Brewing, 120(4), 500-506 (2014)
Dharmendra S Dheeman et al.
Bioresource technology, 102(3), 3373-3379 (2010-12-15)
An extracellular thermostable lipase from Amycolatopsis mediterranei DSM 43304 has been purified to homogeneity using ammonium sulphate precipitation followed by anion exchange chromatography and hydrophobic interaction chromatography. This protocol resulted in a 398-fold purification with 36% final recovery. The purified
Atsushi Ugajin et al.
PloS one, 7(3), e32902-e32902 (2012-03-21)
Anti-predator behaviors are essential to survival for most animals. The neural bases of such behaviors, however, remain largely unknown. Although honeybees commonly use their stingers to counterattack predators, the Japanese honeybee (Apis cerana japonica) uses a different strategy to fight
Hiroshi Sahara et al.
Journal of bioscience and bioengineering, 108(5), 359-364 (2009-10-07)
By application of the high-efficiency loss of heterozygosity (HELOH) method for disrupting genes in diploid sake yeast (Kotaka et al., Appl. Microbiol. Biotechnol., 82, 387-395 (2009)), we constructed, from a heterozygous integrant, a homozygous diploid that overexpresses the alcohol acetyltransferase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service