227471
Amyl acetate, mixture of isomers
Mixture of 2-methylbutyl acetate and n-pentyl acetate, 99%
Synonym(s):
Pentyl acetate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.402
density
0.876 g/mL at 25 °C
functional group
ester
SMILES string
CCCCCOC(C)=O.CCC(C)COC(C)=O
InChI
1S/2C7H14O2/c1-4-6(2)5-9-7(3)8;1-3-4-5-6-9-7(2)8/h6H,4-5H2,1-3H3;3-6H2,1-2H3
InChI key
LSUIXWQPOJYELL-UHFFFAOYSA-N
General description
Amyl acetate is an olfactant. The sensitivity of rat pups towards amyl acetate was studied.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
98.6 °F - closed cup
Flash Point(C)
37 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ontogeny of olfaction: development of the rats' sensitivity to urine and amyl acetate.
Physiology & behavior, 24(5), 965-970 (1980-05-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service