Skip to Content
Merck
All Photos(1)

Key Documents

W355909

Sigma-Aldrich

γ-Terpinene

≥95%, FG

Synonym(s):

1-Isopropyl-4-methyl-1,4-cyclohexadiene, p-Mentha-1,4-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
3559
Beilstein:
2038347
EC Number:
Council of Europe no.:
11025
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.020
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

vapor density

4.7 (vs air)

vapor pressure

~0.7 mmHg ( 20 °C)

Assay

≥95%

refractive index

n20/D 1.474 (lit.)

bp

182 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous; citrus

SMILES string

CC(C)C1=CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,7-8H,5-6H2,1-3H3

InChI key

YKFLAYDHMOASIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

124.9 °F - closed cup

Flash Point(C)

51.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yalda Moayedi et al.
Molecular carcinogenesis, 58(5), 722-734 (2018-12-26)
Bioactive derivatives from the camphor laurel tree, Cinnamomum camphora, are posited to exhibit chemopreventive properties but the efficacy and mechanism of these natural products are not fully understood. We tested an essential-oil derivative, camphor white oil (CWO), for anti-tumor activity
Xiao-Dong Li et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(10), 1544-1548 (2012-03-01)
To extract the essential oils from the Seedlings, the Aseptic Seedlings and the Tissue Culture Seedlings of Thymus vulgaris and analyze their chemical components and the relative contents. The essential oils were extracted by steam distillation, the chemical components and
Yonathan Asikin et al.
Journal of agricultural and food chemistry, 60(32), 7973-7980 (2012-07-19)
Citrus peels are important sources of various pleasant aroma compounds and valuable bioactive substances. To investigate differences in the composition and content of Shiikuwasha (Citrus depressa Hayata) peels from different cultivation lines, the composition of volatile aroma components, flavanones, and
Johanna Rudbäck et al.
Chemical research in toxicology, 25(3), 713-721 (2012-01-19)
The monoterpene α-terpinene is used as a fragrance compound and is present in different essential oils. It is one of the components responsible for the antioxidant activity of tea tree oil. α-Terpinene is structurally similar to other monoterpenes, e.g., limonene
Wei-bin Yuan et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(7), 1067-1069 (2011-11-10)
To analysis the constituents of volatile oil from Fructus Auranti Immaturus by GC-MS. The volatile oil was extracted by steam distillation, then separated by capillary gas chromatography. The constituents of volatile oil were identified and their amount were determined by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service