Skip to Content
Merck
All Photos(1)

Documents

30039

Supelco

p-Cymene

analytical standard

Synonym(s):

1-Isopropyl-4-methylbenzene, 4-Isopropyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH(CH3)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903377
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.62 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)
3.7 mmHg ( 37.7 °C)

Assay

≥99.5% (GC)

autoignition temp.

817 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

5.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.490 (lit.)
n20/D 1.491

bp

176-178 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)c1ccc(C)cc1

InChI

1S/C10H14/c1-8(2)10-6-4-9(3)5-7-10/h4-8H,1-3H3

InChI key

HFPZCAJZSCWRBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: thymus

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rajenahally V Jagadeesh et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(51), 14375-14379 (2011-11-25)
An easily accessible in situ catalyst composed of [{RuCl(2)(p-cymene)}(2)] and terpyridine has been developed for the selective transfer hydrogenation of aromatic nitro and azo compounds. The procedure is general and the selectivity of the catalyst has been demonstrated by applying
Marek Kuzma et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 5174-5182 (2013-02-14)
Asymmetric transfer hydrogenation (ATH) of cyclic imines using [RuCl(η(6)-p-cymene)TsDPEN] (TsDPEN = N-tosyl-1,2-diphenylethylenediamine) was tested with various aliphatic (secondary, tertiary) and aromatic amines employed in the HCOOH-base hydrogen donor mixture. Significant differences in reaction rates and stereoselectivity were observed, which pointed
Gerd Ludwig et al.
Journal of inorganic biochemistry, 113, 77-82 (2012-06-16)
Reactions of ω-diphenylphosphino-functionalized alkyl phenyl sulfides Ph(2)P(CH(2))(n)SPh (n=1, L1; 2, L2; 3, L3), sulfoxides Ph(2)P(CH(2))(n)S(O)Ph (n=1, L4; 2, L5; 3, L6) and sulfones Ph(2)P(CH(2))(n)S(O)(2)Ph (n=1, L7; 2, L8; 3, L9) with the dinuclear chlorido bridged ruthenium(II) complex [{Ru(η(6)-p-cymene)Cl(2)}(2)] afforded mononuclear
Vanessa Kar-Yan Lo et al.
Journal of the American Chemical Society, 134(18), 7588-7591 (2012-04-26)
Complex [(p-cymene)Ru(η(1)-O(2)CCF(3))(2)(OH(2))] mediated transformation of α-diazoacetamides ArCH(2)N(C(CH(3))(3))C(O)CHN(2) to result in carbene insertion into the primary C-H bond exclusively, with the γ-lactam products being isolated in up to 98% yield. This unexpected reaction is striking in view of the presence of
Natalia Busto et al.
Chemistry, an Asian journal, 7(4), 788-801 (2012-02-07)
The reactions of two diaminotriazine ligands 2,4-diamino-6-(2-pyridyl)-1,3,5-triazine (2-pydaT) and 6-phenyl-2,4-diamino-1,3,5-triazine (PhdaT) with ruthenium-arene precursors led to a new family of ruthenium(II) compounds that were spectroscopically characterized. Four of the complexes were cationic, with the general formula [(η(6)-arene)Ru(κ(2)-N,N-2-pydaT)Cl]X (X=BF(4), TsO; arene=p-cymene:

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service