Skip to Content
Merck
All Photos(1)

Key Documents

W276200

Sigma-Aldrich

Myrcene

stabilized, FCC, FG

Synonym(s):

β-Myrcene, 7-Methyl-3-methylene-1,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
136.23
FEMA Number:
2762
Beilstein:
1719990
EC Number:
Council of Europe no.:
2197
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.008
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

vapor density

4.7 (vs air)

vapor pressure

~7 mmHg ( 20 °C)

contains

synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.469 (lit.)

bp

167 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

myrcene

Organoleptic

green; spicy; woody; balsamic; vegetable

storage temp.

2-8°C

SMILES string

C\C(C)=C/CCC(=C)C=C

InChI

1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7H,1,4,6,8H2,2-3H3

InChI key

UAHWPYUMFXYFJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myrcene is one of the key monoterpene volatile compounds of the of Cymbopogon citratus (DC) Stapf leaves[1], mugwort leaves[2] and Xylopia aromatic leaves[3], cotton plant[4], chamchwi plant[5] and guava.[6] It is also a potential nematode attractant.[7]

Application


  • Selective Reactivity of Myrcene for Vat Photopolymerization 3D Printing and Postfabrication Surface Modification.: This research highlights the selective reactivity of myrcene in vat photopolymerization for 3D printing, demonstrating its potential in creating functionalized surfaces for biomedical applications (Weems et al., 2020, Biomacromolecules).

  • Inverse Vulcanization of Sulfur using Natural Dienes as Sustainable Materials for Lithium-Sulfur Batteries.: This article explores the use of myrcene and other natural dienes in the inverse vulcanization of sulfur, promoting sustainable materials for lithium-sulfur batteries (Gomez et al., 2016, ChemSusChem).

Biochem/physiol Actions

Odor at 1%

Other Notes

Natural occurrence: Anise seed, basil, beer, black currant, gin, laurel, rosemary and thyme.

Disclaimer

The substance W276200 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile compounds isolated from edible Korean chamchwi (Aster scaber Thunb).
Chung TY, et al.
Journal of Agricultural and Food Chemistry, 41(10), 1693-1697 (1993)
?-Myrcene, a potent attractant component of pine wood for the Pine Wood Nematode, Bursaphelenchus xylophilus.
Ishkawa M, et al.
Agricultural and Biological Chemistry, 50(7), 1863-1866 (1986)
Analysis of volatile secondary metabolites from Colombian Xylopia aromatica (Lamarck) by different extraction and headspace methods and gas chromatography.
Stashenko EE, et al.
Journal of Chromatography A, 1025(1), 105-113 (2004)
Volatile leaf oil constituents of Cymbopogon citratus (DC) Stapf.
Kasali AA, et al.
Flavour and Fragrance Journal, 16(5), 377-378 (2001)
Volatile flavour components of guava.
Macleod AJ & de Troconis NG.
Phytochemistry, 21(6), 1339-1342 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service