Skip to Content
Merck
All Photos(1)

Key Documents

W278203

Sigma-Aldrich

Nonanal

≥95%, FCC

Synonym(s):

Aldehyde C9, Nonyl aldehyde, Pelargonaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CHO
CAS Number:
Molecular Weight:
142.24
FEMA Number:
2782
Beilstein:
1236701
EC Number:
Council of Europe no.:
114
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.025
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 172.515

vapor pressure

~0.26 mmHg ( 25 °C)

Assay

≥95%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.424 (lit.)

bp

93 °C/23 mmHg (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; fresh; green; citrus; waxy

SMILES string

CCCCCCCCC=O

InChI

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3

InChI key

GYHFUZHODSMOHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonanal is one of the key flavor components of olive oil[1], grapefruit oil[2] and baked potato.[3] It has been reported to be the odorant emitted by humans and birds that attracts Culex mosquitoes.[4]

Application


  • Characteristics of Purified Horse Oil by Supercritical Fluid Extraction with Different Deodorants Agents.: Investigates the efficacy of Nonanal as a deodorizing agent in the purification of horse oil, exploring its potential to improve the sensory qualities of cosmetic products derived from animal fats (Anneke et al., 2024).

  • Characterization of the aroma-active compounds in Xiaokeng green tea by three pretreatment methods combined with gas chromatography-olfactometry (GC-O).: Utilizes Nonanal to identify and characterize the aroma-active compounds in green tea, enhancing understanding of tea flavor chemistry and consumer preferences (Gan et al., 2024).

Biochem/physiol Actions

Taste at 3-10 ppm

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavor components of olive oil?a review.
Kiritsakis AK
Journal of the American Oil Chemists' Society, 75(6), 673-681 (1998)
Effect of cultivar and storage time on the volatile flavor components of baked potato.
Duckham SC, et al.
Journal of Agricultural and Food Chemistry, 50(20), 5640-5648 (2002)
Thomas Van Hecke et al.
PloS one, 9(6), e101122-e101122 (2014-07-01)
The effects of fat content and nitrite-curing of pork were investigated on the formation of cytotoxic and genotoxic lipid oxidation products (malondialdehyde, 4-hydroxy-2-nonenal, volatile simple aldehydes), protein oxidation products (protein carbonyl compounds) and NOC-specific DNA adducts (O6-carboxy-methylguanine) during in vitro
Seung-Ah Lee et al.
Biochimica et biophysica acta, 1782(6), 421-425 (2008-04-09)
Mutations in human Retinol Dehydrogenase 12 (RDH12) are known to cause photoreceptor cell death but the physiological function of RDH12 in photoreceptors remains poorly understood. In vitro, RDH12 recognizes both retinoids and medium-chain aldehydes as substrates. Our previous study suggested
Zainulabeuddin Syed et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(44), 18803-18808 (2009-10-28)
West Nile virus, which is transmitted by Culex mosquitoes while feeding on birds and humans, has emerged as the dominant vector borne disease in North America. We have identified natural compounds from humans and birds, which are detected with extreme

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service