Skip to Content
Merck
All Photos(3)

Key Documents

W263613

Sigma-Aldrich

Linalyl acetate

greener alternative

natural, ≥96%, FG

Synonym(s):

3,7-Dimethyl-1,6-octadien-3-yl acetate, Bergamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
2636
Beilstein:
1724500
EC Number:
Council of Europe no.:
203
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.013
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
biological source:
botanical
food allergen:
no known allergens

biological source

botanical

Quality Level

300
400

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

vapor density

6.8 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

Assay

≥96%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.453 (lit.)

bp

220 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

green; woody; floral; sweet

SMILES string

C\C(C)=C\CCC(C)(OC(C)=O)C=C

InChI

1S/C12H20O2/c1-6-12(5,14-11(4)13)9-7-8-10(2)3/h6,8H,1,7,9H2,2-5H3

InChI key

UWKAYLJWKGQEPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Linalyl acetate is a monoterpene ester mainly found in lavandula essential oil. It is used as a flavoring agent in food industries, as well as a preservative additive in cosmetics and fragrances such as soaps, colognes, perfumes, and skin lotions.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Application


  • Evaluation and estimation of diuretic activity of the linalyl acetate in the rats.: Focuses on the pharmacological effects of linalyl acetate, testing its diuretic activity in rats, which could lead to new therapeutic applications in medicine (Rafique et al., 2024).

  • Characterization of bergamot essential oil: chemical, microbiological and colloidal aspects.: Investigates the components of bergamot essential oil, including linalyl acetate, assessing its chemical properties and potential microbiological uses, supporting its diverse applications in cosmetics and therapeutics (Cordeiro et al., 2024).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essential oil composition and antioxidant activities of eight cultivars of Lavender and Lavandin from western Anatolia
Kivrak S
Industrial Crops and Products, 117, 88-96 (2018)
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 573-573 (2012)
Domenico Trombetta et al.
Antimicrobial agents and chemotherapy, 49(6), 2474-2478 (2005-05-27)
In the present paper, we report the antimicrobial efficacy of three monoterpenes [linalyl acetate, (+)menthol, and thymol] against the gram-positive bacterium Staphylococcus aureus and the gram-negative bacterium Escherichia coli. For a better understanding of their mechanisms of action, the capability
Wafica S Itani et al.
Cancer biology & therapy, 7(11), 1765-1773 (2008-09-13)
Lebanese sage essential oil possesses antitumor properties, however, the bioactive components and antitumor mechanisms are not known. Here we show that combining the three sage bioactive compounds, Linalyl acetate (Ly), Terpeniol (Te) and Camphor (Ca), caused synergistic inhibition of the
Maria Sköld et al.
Contact dermatitis, 58(1), 9-14 (2007-12-25)
Fragrances are among the most common causes of allergic contact dermatitis. We have in previous studies shown that linalool, present in lavender oil, autoxidizes on air exposure, forming allergenic oxidation products. Oxidized linalool was found to be a frequent cause

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service