Skip to Content
Merck
All Photos(1)

Key Documents

W263605

Sigma-Aldrich

Linalyl acetate

≥97%, FCC, FG

Synonym(s):

3,7-Dimethyl-1,6-octadien-3-yl acetate, Bergamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
2636
Beilstein:
1724500
EC Number:
Council of Europe no.:
203
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.013
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor density

6.8 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

description

synthetic

Assay

≥97%

refractive index

n20/D 1.453 (lit.)

bp

220 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; woody; floral; sweet

SMILES string

C\C(C)=C\CCC(C)(OC(C)=O)C=C

InChI

1S/C12H20O2/c1-6-12(5,14-11(4)13)9-7-8-10(2)3/h6,8H,1,7,9H2,2-5H3

InChI key

UWKAYLJWKGQEPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Linalyl acetate is the main component of many plant essential oils such as lavender oil. It can be used as a flavoring agent and fragrance ingredient.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 573-573 (2012)
Maria Sköld et al.
Contact dermatitis, 58(1), 9-14 (2007-12-25)
Fragrances are among the most common causes of allergic contact dermatitis. We have in previous studies shown that linalool, present in lavender oil, autoxidizes on air exposure, forming allergenic oxidation products. Oxidized linalool was found to be a frequent cause
Ali Reza Fakhari et al.
Journal of chromatography. A, 1098(1-2), 14-18 (2005-11-30)
A new method involving concurrent headspace solvent microextraction combined with continuous hydrodistillation (HD-HSME) for the extraction and pre-concentration of the essential oil of Lavandula angustifolia Mill. into a microdrop is developed. A microdrop of n-hexadecane containing n-heptadecane (as internal standard)
C Duclairoir et al.
International journal of pharmaceutics, 253(1-2), 133-144 (2003-02-21)
In this paper, biopolymer nanoparticles are studied, which unlike many synthetic carriers used for controlled release, are biocompatible and biodegradable systems. Gliadins nanoparticles are obtained by a desolvatation method, also known as drawning-out precipitation. These particles have been shown to
Simon Muhoho Njoroge et al.
Journal of agricultural and food chemistry, 51(14), 4029-4035 (2003-06-26)
The chemical changes and artifact formation in daidai (Citrus aurantium L. var. Cyathifera Y. Tanaka) cold-pressed peel oil upon storage at 20, 5, and -21 degrees C for 3, 6, and 12 months were investigated using capillary gas chromatography (GC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service