Skip to Content
Merck
All Photos(1)

Key Documents

W251216

Sigma-Aldrich

Geranyl butyrate

natural, ≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
224.34
FEMA Number:
2512
EC Number:
Council of Europe no.:
274c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.048
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.461 (lit.)

bp

151-153 °C/18 mmHg (lit.)

density

0.896 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; fruity; waxy; rose; sweet

SMILES string

[H]\C(CC\C(C)=C(/[H])COC(=O)CCC)=C(/C)C

InChI

1S/C14H24O2/c1-5-7-14(15)16-11-10-13(4)9-6-8-12(2)3/h8,10H,5-7,9,11H2,1-4H3/b13-10+

InChI key

ZSBOMYJPSRFZAL-JLHYYAGUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geranyl butyrate is one of the key esters that may be contributing to the characteristic flavor of jamun fruit. It also occurs in tomato juice and tomato-based dried products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile flavour components of jamun fruit (Syzygium cumini L).
Vijayanand P, et al.
Flavour and Fragrance Journal, 16(1), 47-49 (2001)
Volatile compounds in tomato-based dried products
Narain N, et al.
Drying Technology, 28(2), 232-239 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service