Skip to Content
Merck
All Photos(1)

Documents

W222208

Sigma-Aldrich

Isobutyric acid

≥99%, FCC, FG

Synonym(s):

Isobutanoic acid, 2-Methylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2H
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2222
Beilstein:
635770
EC Number:
Council of Europe no.:
6
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.006
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.515

vapor density

3.04 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

824 °F

expl. lim.

10 %

refractive index

n20/D 1.393 (lit.)

bp

153-154 °C (lit.)

mp

−47 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

butter; acidic; rancid

SMILES string

CC(C)C(O)=O

InChI

1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)

InChI key

KQNPFQTWMSNSAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyric acid is a volatile fatty acid that can be used as a flavoring agent. It occurs naturally in Ceratonia siliqua L. The gum obtained from the kernels of this species is used as a thickener in the food industry.

Application


  • Isobutyric acid enhances the anti-tumour effect of anti-PD-1 antibody.: This study presents groundbreaking research on how isobutyric acid can enhance the efficacy of anti-PD-1 antibodies, offering significant implications for improving cancer immunotherapy treatments (Murayama et al., 2024).

Biochem/physiol Actions

Odor at 10 ppm
Taste at 15 ppm

Other Notes

Natural occurrence: Roman chamomile, strawberry, dairy products.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manipulation of colonic bacteria and volatile fatty acid production by dietary high amylose maize (amylomaize) starch granules
Wang X, et al
Journal of Applied Microbiology, 93(3), 390-397 (2002)
Effect of temperature on isobutyric acid loss during roasting of carob kibble
Berna A, et al
Journal of Agricultural and Food Chemistry, 45(10), 4084-4087 (1997)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 1463-1464 (1997)
A Vyalikh et al.
Physical chemistry chemical physics : PCCP, 9(18), 2249-2257 (2007-05-10)
Solid state deuterium NMR has been used to study the molecular motion of d(6)-isobutyric acid (d(6)-iBA) in the pure (unconfined) state and confined in the cylindrical pores of two periodic mesoporous silica materials (MCM-41, pore size 3.3 nm and SBA-15
Julia Biermann et al.
Molecular vision, 17, 395-403 (2011-02-12)
Histone deacetylase inhibitors (HDACi) have neuroprotective effects under various neurodegenerative conditions, e.g., after optic nerve crush (ONC). HDACi-mediated protection of central neurons by increased histone acetylation has not previously been demonstrated in rat retinal ganglion cells (RGCs), although epigenetic changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service