Skip to Content
Merck
All Photos(1)

Key Documents

108154

Sigma-Aldrich

Hexyl acetate

99%

Synonym(s):

Capryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)5CH3
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747138
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

refractive index

n20/D 1.409 (lit.)

bp

168-170 °C (lit.)

mp

−80 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCCCOC(C)=O

InChI

1S/C8H16O2/c1-3-4-5-6-7-10-8(2)9/h3-7H2,1-2H3

InChI key

AOGQPLXWSUTHQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexyl acetate is an ester and is commonly used as a solvent for resins, polymers, fats, and oils. It can also be used as a flavoring agent in the food industry.[1] It is produced by acid catalyzed liquid phase esterification of n-hexanol and acetic acid[2].

Application

Hexyl acetate was used to study the activity of diamondback moth sex pheromone and larval frass volatiles, as well as green leaf volatiles from cabbage, on the natural enemies of the pest[3].

Biochem/physiol Actions

Hexyl acetate has antimicrobial activity and can be used to improve the safety of minimally processed fruits[4]. Hexyl acetate is a fruity smelling fluid used as flavoring agent or in perfumes[2]. Hexyl acetate is a green leaf volatile from cabbage Brassica oleracea var. capitata L[3].

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intensification of enzyme catalysed synthesis of hexyl acetate using sonication
AR Deshmukh, et al.
Green Processing and Synthesis, 6, 55-62 (2017)
G V P Reddy et al.
Journal of chemical ecology, 28(1), 131-143 (2002-03-02)
The parasitoids Trichogramma chilonis (Hymenoptera: Trichogrammatidae) and Cotesia plutellae (Hymenoptera: Braconidae), and the predator Chrysoperla carnea (Neuroptera: Chrysopidae), are potential biological control agents for the diamondback moth, Plutella xylostella (Lepidoptera: Yponomeutidae). We present studies on the interactions between these bioagents
Rosalba Lanciotti et al.
Journal of agricultural and food chemistry, 51(10), 2958-2963 (2003-05-02)
The aims of this work were to evaluate the effects of different concentrations of hexanal, (E)-2-hexenal, hexyl acetate, and their mixtures on the fate of pathogenic species such as Escherichia coli, Salmonella enteritidis, and Listeria monocytogenes inoculated in model systems
Sven R L Gobert et al.
Journal of microencapsulation, 36(4), 371-384 (2019-06-20)
Microencapsulation is almost exclusively performed in batch processes. With today's chemistry increasingly performed in flow reactors, this work aims to realise a continuous reactor setup for the encapsulation of an ester with a polyuria (PU) shell. The generation of an
Synthesis of n-hexyl acetate by reactive distillation.
Schmitt M, et al.
Chemical Engineering and Processing, 43(3), 397-409 (2004)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service