T60402
Triethyl orthoacetate
97%
Synonym(s):
1,1,1-Triethoxyethane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C(OC2H5)3
CAS Number:
Molecular Weight:
162.23
Beilstein:
506201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.396 (lit.)
bp
142 °C (lit.)
density
0.885 g/mL at 25 °C (lit.)
SMILES string
CCOC(C)(OCC)OCC
InChI
1S/C8H18O3/c1-5-9-8(4,10-6-2)11-7-3/h5-7H2,1-4H3
InChI key
NDQXKKFRNOPRDW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Triethyl orthoacetate can be used for the synthesis of:
- Ordered mesoporous carbons for catalysis, electrochemistry and hydrogen storage applications.
- Esters of phosphonic acids, carboxylic acids and phosphinic acids.
- Ethyl alk-4-enoates by Johnson–Claisen rearrangement reaction.
- Benzoxazoles, benzimidazoles and oxazolo[4,5-b]pyridines.
- Quinazolin-4(3H)-one derivatives.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
102.2 °F - Non-equilibrium method
Flash Point(C)
39 °C - Non-equilibrium method
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Silica sulfuric acid catalyzed synthesis of benzoxazoles, benzimidazoles and oxazolo [4, 5-b] pyridines under heterogeneous and solvent-free conditions
Mohammadpoor-Baltork I, et al.
Journal of the Iranian Chemical Society, 5(1), S65-S70 (2008)
Synthesis of ordered mesoporous carbons with channel structure from an organic-organic nanocomposite
Tanaka S, et al.
Chemical Communications (Cambridge, England), 16, 2125-2127 (2005)
Efficient esterification of carboxylic acids and phosphonic acids with trialkyl orthoacetate in ionic liquid
Yoshino T, et al.
Tetrahedron, 62(6), 1309-1317 (2006)
Treatment of Baylis-Hillman adducts with triethyl orthoacetate in the presence of heterogeneous catalysts: a method for the stereoselective synthesis of two different types of trisubstituted alkenes
Das B, et al.
Tetrahedron Letters, 47(43), 7619-7623 (2006)
A new approach to the facile synthesis of mono-and disubstituted quinazolin-4 (3H)-ones under solvent-free conditions.
Salehi P, et al.
Tetrahedron Letters, 46(41), 7051-7053 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service