Skip to Content
Merck
All Photos(2)

Documents

333344

Sigma-Aldrich

Tripropyl orthoformate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH(OCH2CH2CH3)3
CAS Number:
Molecular Weight:
190.28
Beilstein:
1744249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.407 (lit.)

bp

106-108 °C/40 mmHg (lit.)

density

0.883 g/mL at 25 °C (lit.)

SMILES string

CCCOC(OCCC)OCCC

InChI

1S/C10H22O3/c1-4-7-11-10(12-8-5-2)13-9-6-3/h10H,4-9H2,1-3H3

InChI key

RWNXXQFJBALKAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Gas phase reactions of the rare earth metal cations with tripropyl orthoformate were studied by Fourier transform ion cyclotron resonance mass spectrometry.

Application

Tripropyl orthoformate was used as alcohol donor to investigate the activity and enantioselectivity of Novozym 435 in the esterification of rac-fenoprofen in methylcyclohexane. It may be used as a drying agent during the synthesis of squaraine derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of squaraine dyes under mild conditions: applications for labelling and sensing of biomolecules.
Sleiman MH and Ladame S.
Chemical Communications (Cambridge, England), 50(40), 5288-5290 (2014)
The structure/property relationship in a series of pyrrolic polysquaraines.
Lynch DE and Ewington J.
Synt. Metals, 183, 40-49 (2013)
Advances in synthesis and application of near-infrared absorbing squaraine dyes.
Hu L, et al.
Royal Society of Chemistry Advances, 3(21), 7667-7676 (2013)
Gas phase reactivity of rare earth metal cations with trialkylorthoformates: synthesis of neutral rare earth alkoxides.
Marchande N, et al
International Journal of Mass Spectrometry, 195, 139-148 (2000)
Convenient preparation of (s)-fenoprofen by biocatalysed irreversible esterification.
Morrone R, et al.
Rasayan Journal of Chemistry, 1, 732-737 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service