T60453
Triethyl orthoformate
reagent grade, 98%
Synonym(s):
1,1,1-Triethoxymethane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH(OC2H5)3
CAS Number:
Molecular Weight:
148.20
Beilstein:
605384
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
Assay:
98%
bp:
146 °C (lit.)
vapor pressure:
2.9 mmHg ( 20 °C)
Recommended Products
grade
reagent grade
Quality Level
vapor density
5.11 (vs air)
vapor pressure
2.9 mmHg ( 20 °C)
Assay
98%
form
liquid
refractive index
n20/D 1.391 (lit.)
bp
146 °C (lit.)
mp
−76 °C (lit.)
density
0.891 g/mL at 25 °C (lit.)
SMILES string
CCOC(OCC)OCC
InChI
1S/C7H16O3/c1-4-8-7(9-5-2)10-6-3/h7H,4-6H2,1-3H3
InChI key
GKASDNZWUGIAMG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Triethylorthoformate is an organic compound that is soluble in most organic solvents. It is commonly used as a solvent for thallium trinitrate oxidation reactions.
Application
Triethyl orthoformate can be used as a:
- Precursor for the controlled release of ethylformate(EF) vapor.
- Solvent in the experimental determination of the solubility of ε-CL-20 at different temperatures (ranging from 278.15 to 318.15K) and a constant pressure of 0.1 MPa.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
95.0 °F - closed cup
Flash Point(C)
35 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Triethyl orthoformate
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
In-package fumigation of blueberries using ethyl formate: Effects on spotted-wing drosophila (Drosophila suzukii Matsumura) mortality and fruit quality
Zaitoon A, et al.
Food Packaging and Shelf Life, 30, 100717-100717 (2021)
Measurement and Correlation of the Solubility of $\varepsilon$-CL-20 in 12 Organic Solvents at Temperatures Ranging from 278.15 to 318.15 K
Lv P, et al.
Journal of Chemical and Engineering Data, 62(3), 961-966 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service