P66807
3-Pyridinemethanol
98%
Synonym(s):
ω-Hydroxy-3-picoline, 3-(Hydroxymethyl)pyridine, 3-Pyridyl carbinol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
107851
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.545 (lit.)
bp
154 °C/28 mmHg (lit.)
density
1.124 g/mL at 25 °C (lit.)
SMILES string
OCc1cccnc1
InChI
1S/C6H7NO/c8-5-6-2-1-3-7-4-6/h1-4,8H,5H2
InChI key
MVQVNTPHUGQQHK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
3-Pyridinemethanol, an aromatic primary alcohol, is the key moiety of many bio-active and industrially important compounds.
Application
3-Pyridinemethanol can undergo aerobic photo-oxidation in the presence of catalytic amount of hydrobromic acid to form nicotinic acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
275.0 °F - closed cup
Flash Point(C)
135 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Conformational stability, molecular structure, vibrational, electronic, 1H and 13C spectral analysis of 3-pyridinemethanol using ab-initio/DFT method.
Sivaranjani T
Journal of Molecular Structure, 1108, 398-410 (2016)
Aerobic photo-oxidation of alcohols in the presence of a catalytic inorganic bromo source.
Hirashima S
Tetrahedron, 62(33), 7887-7891 (2006)
T Fazekas et al.
Acta physiologica Hungarica, 74(2), 169-174 (1989-01-01)
The antilipolytic, nicotinic acid analogue beta-pyridylcarbinol (Ronicol) has previously been reported to decrease the free fatty acid (FFA) concentration of the arteria-blood, and to moderate the FFA-uptake and O2-consumption of the myocardium; on this basis, the drug may be expected
C Keller et al.
Klinische Wochenschrift, 66(14), 647-650 (1988-07-15)
We report two cases of unusual hepatic lesions following years of monotherapy with pyridylcarbinol (timespan) in a daily dose of 1.35 to 1.5 g for familial hypercholesterolemia. Mild elevations of liver enzymes correlated to a large tumor-like mass in the
T B Gelhard et al.
Journal de biologie buccale, 16(2), 95-99 (1988-06-01)
The F- uptake in vivo in sound human enamel from fluoridated toothpastes is described. Ten participants brushed for about 1 min twice a day, four enamel specimens in a dental appliance over three week periods. The enamel specimens were prepared
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service