Skip to Content
Merck
All Photos(1)

Key Documents

H57009

Sigma-Aldrich

3-Hydroxypyridine

98%

Synonym(s):

3-Pyridinol, 3-Pyridone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
Beilstein:
105699
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

125-128 °C (lit.)

SMILES string

Oc1cccnc1

InChI

1S/C5H5NO/c7-5-2-1-3-6-4-5/h1-4,7H

InChI key

GRFNBEZIAWKNCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fazlul Huq et al.
European journal of medicinal chemistry, 39(8), 691-697 (2004-07-28)
Four trans-planaramineplatinum(II) complexes code named YH9, YH10, YH11 and YH12, each of the form trans-PtL(NH(3))Cl(2) where L = 2-hydroxypyridine and 3-hydroxypyridine, imidazole, and imidazo(1,2-alpha)pyridine for YH9, YH10, YH11 and YH12, respectively. All of the compounds have significant anticancer activity against
G S M Kiruba et al.
The Journal of organic chemistry, 68(7), 2874-2881 (2003-03-29)
The tautomeric equilibria of a series of 3-hydroxypyridine derivatives including pyridoxal-5'-phosphate (PLP), the active form of vitamin B(6), have been studied using density functional calculations (B3LYP/6-311+G//B3LYP/6-31G) in the gas phase and in different solvents. Three different approaches, namely continuum, discrete
L D Lukyanova et al.
Bulletin of experimental biology and medicine, 148(4), 587-591 (2010-04-17)
Succinate-containing derivatives of 3-hydroxypyridine, mexidol and proxypin, serve as succinate donors for the respiratory chain and contribute to activation of the succinate oxidase pathway of oxidation. Under conditions of hypoxia, these changes promote recovery of aerobic energy production, normalization of
Zhongfa Liu et al.
Chemical research in toxicology, 16(2), 232-241 (2003-02-18)
2-hydroxyaldehydes have been previously identified as products of lipid peroxidation, and although they represent the simplest reducing sugars, their potential for modification of proteins under physiological conditions has not been investigated. Here, 2-hydroxyaldehydes were found to condense with amines in
Osama K Abou-Zied et al.
Journal of the American Chemical Society, 130(32), 10793-10801 (2008-07-23)
Subdomain IIA binding site of human serum albumin (HSA) was characterized by examining the change in HSA fluorescence in the native, unfolded, and refolded states. The study was carried out in the absence and presence of small molecular probes using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service