Skip to Content
Merck
All Photos(2)

Key Documents

O5500

Sigma-Aldrich

Octyl acetate

≥99%

Synonym(s):

Acetic acid octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2(CH2)7CH3
CAS Number:
Molecular Weight:
172.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.9 (vs air)

Assay

≥99%

autoignition temp.

515 °F

expl. lim.

8.14 %

refractive index

n20/D 1.418 (lit.)

bp

211 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCOC(C)=O

InChI

1S/C10H20O2/c1-3-4-5-6-7-8-9-12-10(2)11/h3-9H2,1-2H3

InChI key

YLYBTZIQSIBWLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

189.1 °F - closed cup

Flash Point(C)

87.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J A de Bruijn et al.
Journal of evolutionary biology, 29(3), 665-671 (2015-12-22)
Animals often respond to danger by raising alarm to inform others. Alarm signals come in many different forms, such as visual or mechanical display, sound or odour. Some animals produce vocal alarm signals that vary with the level of danger.
Pachara Sattayawat et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(3), 1404-1413 (2020-01-10)
Bio-based production technologies may complement or replace petroleum-based production of chemicals, but they face a number of technical challenges, including product toxicity and/or water insolubility. Plants and microorganisms naturally biosynthesize chemicals that often are converted into derivatives with reduced toxicity
W C Daughtrey et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 12(2), 313-320 (1989-02-01)
The subchronic toxicity of octyl acetate was assessed following its administration to rats via oral gavage, 5 days per week for 13 weeks. Treated rats received undiluted octyl acetate at doses of 0.1, 0.5, or 1.0 g/kg. Control rats received
C Ursin et al.
American Industrial Hygiene Association journal, 56(7), 651-660 (1995-07-01)
A procedure has been developed for measuring the steady state rate of permeation of commercial solvents through living human skin. To get the most consistent results, it was necessary with some solvents to normalize the solvent permeation rate of a
Xue Zhou et al.
Mycologia, 110(4), 710-725 (2018-09-06)
Fusarium zanthoxyli and F. continuum are sister taxa that are the etiological agents of canker disease of prickly ash (Zanthoxylum bungeanum) in northern China. These two pathogens, together with F. torreyae, the causal agent of canker disease of the critically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service