Skip to Content
Merck
All Photos(2)

Documents

108731

Sigma-Aldrich

2-Nonanone

≥99%

Synonym(s):

Heptyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COCH3
CAS Number:
Molecular Weight:
142.24
Beilstein:
1743645
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021224
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

refractive index

n20/D 1.421 (lit.)

bp

192 °C/743 mmHg (lit.)

mp

−21 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC(C)=O

InChI

1S/C9H18O/c1-3-4-5-6-7-8-9(2)10/h3-8H2,1-2H3

InChI key

VKCYHJWLYTUGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Nonanone is a repulsive odorant and can be detected by the AWB olfactory neurons. It is a medium toxic compound with a rat LD50 of 400–4000 mgkg-1. It is an effective antifungal agent.

Application

2-Nonanone was used to analyze the sensitivity of sensors coated with nafion and carbowax to certify cheese samples. It was used to study binding of β-lactoglobulin to alkanones.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Control of Decay in Modified-atmosphere Packages of Sliced Apples using 2-Nonanone Vapor.
Leepipattanawit R, et al.
Journal of Food Science, 62(5), 1043-1047 (1997)
Binding of alkanone flavors to β-lactoglobulin: effects of conformational and chemical modification.
O'Neill TE and Kinsella JE.
Journal of Agricultural and Food Chemistry, 35(5), 770-774 (1987)
The neurotoxicity of 5-nonanone: preliminary report.
M A Shifman et al.
Toxicology letters, 8(4-5), 283-288 (1981-06-01)
E R Troemel et al.
Cell, 91(2), 161-169 (1997-11-05)
Different olfactory cues elicit distinct behaviors such as attraction, avoidance, feeding, or mating. In the nematode C. elegans, these cues are sensed by a small number of olfactory neurons, each of which expresses several different odorant receptors. The type of
D A Birnby et al.
Genetics, 155(1), 85-104 (2000-05-03)
Caenorhabditis elegans daf-11 and daf-21 mutants share defects in specific chemosensory responses mediated by several classes of sensory neurons, indicating that these two genes have closely related functions in an assortment of chemosensory pathways. We report that daf-11 encodes one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service