Skip to Content
Merck
All Photos(1)

Documents

M29606

Sigma-Aldrich

9-Methylanthracene

98%

Synonym(s):

9-Methylanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
Beilstein:
1907463
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

196-197 °C/12 mmHg (lit.)

mp

77-79 °C (lit.)

density

1.066 g/mL at 25 °C (lit.)

SMILES string

Cc1c2ccccc2cc3ccccc13

InChI

1S/C15H12/c1-11-14-8-4-2-6-12(14)10-13-7-3-5-9-15(11)13/h2-10H,1H3

InChI key

CPGPAVAKSZHMBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wenyan Shi et al.
Polymers, 11(3) (2019-04-10)
Membrane inlet mass spectrometry (MIMS) is commonly used for detecting the components in liquid samples. When a liquid sample flows through a membrane, certain analytes will permeate into the vacuum chamber of a mass spectrometer from the solution. The properties
Kamlesh Awasthi et al.
The journal of physical chemistry. A, 113(40), 10603-10609 (2009-10-02)
Photoluminescence of electron donor-acceptor pairs that show photoinduced electron transfer (PIET) has been measured in a polymer film under simultaneous application of electric field and magnetic field. Fluorescence emitted from the locally excited state (LE fluorescence) of 9-methylanthracene (MAnt) and
Samantha L Jenkins et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(4-5), 1131-1139 (2005-06-15)
Reaction of single crystals of benzoic and trans-cinnamic acids with 200 Torr pressure of ammonia gas in a sealed glass bulb at 20 degrees C generates the corresponding ammonium salts; there is no sign of any 1:2 adduct as has
Klaus Pittertschatscher et al.
Analytical biochemistry, 308(2), 300-306 (2002-11-07)
A novel one-step ethylchloroformate (ECF) derivatization of histamine in biological liquid matrices that allows the sensitive quantification by gas chromatography and mass spectroscopic detection (GC-MS) from small volumes of blood plasma or cell culture supernatants within 15 min is described.
K Takegoshi et al.
Solid state nuclear magnetic resonance, 11(3-4), 189-196 (1998-08-07)
The locus of the photodimerization reaction of 9-methylanthracene in the crystal was examined by high-resolution solid-state 13C NMR techniques. Examination of the 13C spectra of the products showed that only the trans dimer is formed by the solid state photodimerization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service