Skip to Content
Merck
All Photos(1)

Documents

35754

Supelco

Chrysene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909297
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

448 °C (lit.)

mp

252-254 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2c(c1)ccc3c4ccccc4ccc23

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H

InChI key

WDECIBYCCFPHNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chrysene is a symmetrical polycyclic aromatic hydrocarbon (PAH) with four condensed benzene rings. Incomplete combustion of fossil fuels and other industrial processes results in the production of chrysene which is known to be toxic, mutagenic and carcinogenic to humans.

Application

Chrysene may be used as an analytical reference standard for the determination of the analyte in fish bile, air particulate extracts and food samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of PAH in food samples by HPLC with fluorimetric detection following sonication extraction without sample clean-up.
Nieva-Cano MJ, et al.
Analyst, 126(8), 1326-1331 (2001)
Quantitative determination of de-conjugated chrysene metabolites in fish bile by HPLC-fluorescence and GC-MS.
Jonsson G, et al.
Chemosphere, 54(8) (2004)
Liquid chromatographic determination of polycyclic aromatic hydrocarbons in air particulate extracts.
May WE and Wise SA
Analytical Chemistry, 56(2), 225-232 (1984)
Biodegradation of chrysene, an aromatic hydrocarbon by Polyporus sp. S133 in liquid medium.
Hadibarata T, et al.
Journal of Hazardous Materials, 164(2-3), 911-917 (2009)
Peter A Lieberzeit et al.
Analytical and bioanalytical chemistry, 392(7-8), 1405-1410 (2008-11-06)
Molecular imprinting with binary mixtures of different polycyclic aromatic hydrocarbons (PAH) is a tool for design of chemically highly sensitive layers for detection of these analytes. Sensor responses increase by one order of magnitude compared with layers imprinted with one

Articles

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service