Product Description
Zinc, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.9%
Zinc, foil, 8mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 4mm disks, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, not light tested, 25x25mm, thickness 0.015mm, 99.9%
Zinc, foil, 2m coil, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 5m coil, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 8mm disks, thickness 0.01mm, 99.9%
Zinc, foil, not light tested, 50x50mm, thickness 0.015mm, 99.9%
Zinc, foil, 50mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 100x100mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95+%