Skip to Content
Merck
All Photos(4)

Documents

D222208

Sigma-Aldrich

Dodecylamine

98%

Synonym(s):

1-Aminododecane, Laurylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11NH2
CAS Number:
Molecular Weight:
185.35
Beilstein:
1633576
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

64 mmHg ( 170 °C)

Quality Level

Assay

98%

form

crystals

bp

247-249 °C (lit.)

mp

27-29 °C (lit.)

density

0.806 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCN

InChI

1S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3

InChI key

JRBPAEWTRLWTQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dodecylamine (DDA) can be used:
  • As a modifier in the preparation of dodecylamine incorporated sodium montmorillonite. It is used as an adsorbent for hexavalent chromium.
  • In the synthesis of DDA-poly(aspartic acid) as a biodegradable water-soluble polymeric material.
  • As an organic surfactant in the synthesis of Sn(IV)-containing layered double hydroxide (LDHs), which can be further used as ion exchangers, absorbents, ion conductors, and catalysts.
  • As a complexing, reducing and capping agent in the synthesis of pentagonal silver nanowires.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract,Liver,Immune system, Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and catalytic properties of Sn-containing layered double hydroxides synthesized in the presence of dodecylsulfate and dodecylamine
Tong DS, et al.
Applied Clay Science, 48(4), 569-574 (2010)
Potential application of dodecylamine modified sodium montmorillonite as an effective adsorbent for hexavalent chromium
Kumar ASK, et al.
Chemical Engineering Journal, 211, 396-405 (2012)
Zheng Xing et al.
Physical chemistry chemical physics : PCCP, 19(32), 21426-21435 (2017-08-02)
This work investigates the effect of atmosphere on pyrolysis of a polymer matrix (precursor) for directing its transformation towards more disordered graphene species and smaller graphitic nanograins. These two structural characteristics are crucial to the generation of nano-channels (NCs) pertinent
Preparation and characterization of dodecylamine-modified poly (aspartic acid) as a biodegradable water-soluble polymeric material
Nakato T, et al.
Polymer Bull., 44(4), 385-391 (2000)
Controllable synthesis of pentagonal silver nanowires via a simple alcohol-thermal method
You T, et al.
Materials Letters, 63(11), 920-922 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service