Skip to Content
Merck
All Photos(1)

Key Documents

305391

Sigma-Aldrich

Octadecylamine

≥99% (GC)

Synonym(s):

1-Aminooctadecane, Stearylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17NH2
CAS Number:
Molecular Weight:
269.51
Beilstein:
636111
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

10 mmHg ( 72 °C)

Quality Level

Assay

≥99% (GC)

bp

232 °C/32 mmHg (lit.)

mp

50-52 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCN

InChI

1S/C18H39N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-19H2,1H3

InChI key

REYJJPSVUYRZGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octadecylamine (ODA) is a primary alkyl amine mainly used as a hydrophobic surface modifier.[1]

Application

Octadecylamine is used to surface functionalize different carbon nanomaterials (graphene oxide,[2] carbon nanotubes) for different applications which include thin film nanocomposite (TFN) nanofilteration[3] and carbon fiber microelectrodes.[4] It can be used for the preparation of butyrylcholinesterase/stearylamine films (Langmuir-Blodgett films) for use in enzymatic field effect transistor (ENFET) based biosensors.[5] Octadecylamine also forms films which can be used in ion exachnge systems.[6] It may also be used in the preparation of metal oxide nano crystals with controlled size and shape.[7]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Target Organs

Liver,Gastrointestinal tract,Immune system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Octadecylamine-based Langmuir-Blodgett films containing iron and copper hexacyanoferrates.
Ivanov NS, et al.
Colloid Journal, 77(2), 154-159 (2015)
Chlorine resistant TFN nanofiltration membrane incorporated with octadecylamine-grafted GO and fluorine-containing monomer.
Xue S-M, et al.
Journal of Membrane Science , 545(14), 185-195 (2018)
Enzyme-Octadecylamine Langmuir-Blodgett membranes for ENFET biosensors
Wan K, et al.
Talanta, 52(4), 663-670 (2000)
Fabrication of a novel octadecylamine functionalized graphene oxide/PVDF dual-layer flat sheet membrane for desalination via air gap membrane distillation.
Zahirifar J, et al.
Desalination, 428(14), 227-239 (2018)
Functional groups modulate the sensitivity and electron transfer kinetics of neurochemicals at carbon nanotube modified microelectrodes.
Jacobs CB, et al.
Analyst, 136(17), 3557-3565 (2011)

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service