Skip to Content
Merck
All Photos(1)

Documents

O1408

Sigma-Aldrich

Octadecylamine

technical grade, 90%

Synonym(s):

1-Aminooctadecane, Stearylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17NH2
CAS Number:
Molecular Weight:
269.51
Beilstein:
636111
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor pressure

10 mmHg ( 72 °C)

Assay

90%

bp

232 °C/32 mmHg (lit.)

mp

50-52 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCN

InChI

1S/C18H39N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-19H2,1H3

InChI key

REYJJPSVUYRZGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Target Organs

Liver,Gastrointestinal tract,Immune system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong-Kai Cui et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13668-13674 (2012-08-31)
It has been shown that mixtures of monoalkylated amphiphiles and sterols can form liquid-ordered (lo) lamellar phases. These bilayers can be extruded using conventional methods to obtain large unilamellar vesicles (LUVs) that have very low permeability and a specific response
Cui Mao et al.
ACS applied materials & interfaces, 4(10), 5281-5286 (2012-09-07)
Polystyrene (PS) and poly(methyl methacrylate) (PMMA) blends filled with octadecylamine-functionalized graphene (GE-ODA) have been fabricated to obtain conductive composites with a lower electrical percolation threshold according to the concept of double percolation. The dependence of the electrical properties of the
Cristiane Moraes Santos et al.
Journal of biomedical nanotechnology, 8(2), 322-329 (2012-04-21)
Topical treatment of cutaneous leishmaniasis represents an exciting alternative for reducing toxicity associated with parenteral administration of conventional amphotericin B. This work aims to develop and to characterize amphotericin B-loaded new carriers and to investigate their potential for topical delivery
Kandadi Prabhakar et al.
Colloids and surfaces. B, Biointerfaces, 86(2), 305-313 (2011-05-14)
Indinavir, an antiretroviral protease inhibitor used in treatment of HIV infection has limited penetration into brain due to efflux of P-glycoprotein. The aim of this work was to develop transferrin coupled submicron lipid emulsions (SLEs) containing indinavir for delivery to
D Pandita et al.
Die Pharmazie, 66(3), 171-177 (2011-05-11)
The objective of the study was to design and evaluate a solid lipid nanoparticle (SLN) drug delivery system for delivery of paclitaxel. Components of the SLN were lipid (stearylamine) and surfactants (Pluronic F68 and Soya lecithin). The paclitaxel loaded nanoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service