Skip to Content
Merck
All Photos(3)

Key Documents

C94009

Sigma-Aldrich

4-Cyanophenol

95%

Synonym(s):

4-Hydroxybenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4OH
CAS Number:
Molecular Weight:
119.12
Beilstein:
386130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

crystals

mp

110-113 °C (lit.)

SMILES string

Oc1ccc(cc1)C#N

InChI

1S/C7H5NO/c8-5-6-1-3-7(9)4-2-6/h1-4,9H

InChI key

CVNOWLNNPYYEOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Cyanophenol is a precursor for the synthesis of a vasodilator, Levcromakalim. Bromination of 4-cyanophenol results in bromoxynil, a commercial herbicide. It can also be used as a component of deep eutectic solvent (DES) mixture.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High atom efficient and environment-friendly preparation of herbicides bromoxynil and ioxynil.
Joshi G and Patil RD
Indian Journal of Chemistry, 49B, 1678-1680 (2010)
Synthesis and antihypertensive activity of 4-(cyclic amido)-2H-1-benzopyrans.
Ashwood VA, et al.
Journal of Medicinal Chemistry, 29(11), 2194-2201 (1986)
Nitrogen-doped carbons prepared from eutectic mixtures as metal-free oxygen reduction catalysts.
Lopez-Salas N, et al.
Journal of Material Chemistry A, 4(2), 478-488 (2016)
D B Harper
The International journal of biochemistry, 17(6), 677-683 (1985-01-01)
The purification and properties of an enzyme from Nocardia sp. which catalyses the conversion of p-hydroxybenzonitrile to p-hydroxybenzoic acid and ammonia without intermediate formation of the amide is described. The enzyme displayed a broad pH optimum between 7.0 and 9.5
J E Harrison et al.
Pharmaceutical research, 13(4), 542-546 (1996-04-01)
The purpose of this work was to analyse the mechanism of the enhancement of percutaneous penetration demonstrated by the known enhancers Azone and Transcutol. Enhancer induced changes in the diffusivity and solubility of a model permeant (4-cyanophenol) in human stratum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service